This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05015591_s1
See other TMEM245 GE Assays ›
Gene Symbol:
TMEM245
Gene Name:
transmembrane protein 245
Gene Aliases:
C9orf5, CG-2, CG2
Chromosome Location:
Chr.9: 109015135 - 109119963 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05015591_s1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMEM245
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_032012.3 NP_114401.2 18 3781 130
XM_011518446.2 XP_011516748.1 18 3796 130
XM_011518447.1 XP_011516749.1 17 3757 130
XM_011518449.1 XP_011516751.1 17 3664 130
XM_011518451.1 XP_011516753.1 16 3652 130
XM_011518452.1 XP_011516754.1 17 3547 130
XM_017014571.1 XP_016870060.1 17 3754 130
XM_017014572.1 XP_016870061.1 16 3523 130
GenBank mRNA AF086175.1 - 1 130
AF153415.1 - 18 3786 130
AK074248.1 - 8 2079 130
AK090543.1 - 4 2836 130
BC014248.2 - 8 2132 130

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TMEM245

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000472207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000472207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000472207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000472207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000472207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000472207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000472207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000472207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000472207.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000482868.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000482868.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000482868.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000482868.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000374586.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000413712.6 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000472207.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000472207.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000472207.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000472207.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000472207.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000472207.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000472207.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000472207.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000472207.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000482868.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000482868.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000482868.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000482868.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 ENST00000491854.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Clariom D Assay TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Clariom D Assay TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Clariom D Assay TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Clariom D Assay TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Clariom D Assay TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Clariom D Assay TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Clariom D Assay TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Clariom D Assay TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Clariom D Assay TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Clariom D Assay TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Clariom D Assay TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Clariom D Assay TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Clariom D Assay TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Clariom D Assay TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Clariom D Assay TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Clariom D Assay TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Clariom D Assay TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Clariom D Assay TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Clariom D Assay TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Clariom D Assay TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Clariom D Assay TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Clariom D Assay TC0900011151.hg.1 NM_032012.3 Refseq
Affymetrix Human Clariom D Assay TC0900011151.hg.1 NM_032012.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0