This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05011838_s1
See other ROR1 GE Assays ›
Gene Symbol:
ROR1
Gene Name:
receptor tyrosine kinase like orphan receptor 1
Gene Aliases:
NTRKR1, dJ537F10.1
Chromosome Location:
Chr.1: 63774019 - 64181498 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05011838_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ROR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005012.3 NP_005003.2 9 4200 73
XM_011541526.1 XP_011539828.1 9 3799 73
XM_017001376.1 XP_016856865.1 8 4100 73
XM_017001377.1 XP_016856866.1 11 6743 73
GenBank mRNA AK000776.1 - 1 218 73

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ROR1

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

transmembrane receptor protein tyrosine kinase signaling pathway
peptidyl-tyrosine phosphorylation
Wnt signaling pathway, planar cell polarity pathway
transmembrane receptor protein tyrosine kinase activity
protein binding
ATP binding
Wnt-protein binding
coreceptor activity involved in Wnt signaling pathway, planar cell polarity pathway

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000371079.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000371079.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000371079.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000371079.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000371079.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000371079.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000371079.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000371079.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000371079.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000371079.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000371079.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000371079.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000371079.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000371080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000371080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000371080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000371080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000371080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000371080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000371080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000371080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000371080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000371080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000482426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000482426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000482426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000482426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000482426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000482426.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000545203.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000545203.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000545203.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000545203.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000545203.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000545203.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000545203.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000545203.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000545203.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000545203.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 ENST00000545203.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 NM_001083592.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 NM_001083592.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 NM_001083592.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 NM_001083592.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 NM_001083592.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 NM_001083592.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 NM_001083592.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 NM_001083592.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 NM_001083592.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 NM_001083592.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 NM_005012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 NM_005012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 NM_005012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 NM_005012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 NM_005012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 NM_005012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 NM_005012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 NM_005012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 NM_005012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 NM_005012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 NM_005012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 NM_005012.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008594.hg.1 NM_005012.3 Refseq
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000371079.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000371079.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000371079.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000371079.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000371079.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000371079.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000371079.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000371079.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000371079.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000371079.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000371079.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000371079.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000371079.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000371080.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000371080.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000371080.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000371080.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000371080.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000371080.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000371080.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000371080.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000371080.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000371080.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000482426.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000482426.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000482426.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000482426.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000482426.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000482426.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000545203.2 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000545203.2 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000545203.2 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000545203.2 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000545203.2 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000545203.2 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000545203.2 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000545203.2 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000545203.2 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000545203.2 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 ENST00000545203.2 Ensembl
Affymetrix Human Clariom D Assay TC0100008594.hg.1 NM_001083592.1 Refseq
Affymetrix Human Clariom D Assay TC0100008594.hg.1 NM_001083592.1 Refseq
Affymetrix Human Clariom D Assay TC0100008594.hg.1 NM_001083592.1 Refseq
Affymetrix Human Clariom D Assay TC0100008594.hg.1 NM_001083592.1 Refseq
Affymetrix Human Clariom D Assay TC0100008594.hg.1 NM_001083592.1 Refseq
Affymetrix Human Clariom D Assay TC0100008594.hg.1 NM_001083592.1 Refseq
Affymetrix Human Clariom D Assay TC0100008594.hg.1 NM_001083592.1 Refseq
Affymetrix Human Clariom D Assay TC0100008594.hg.1 NM_001083592.1 Refseq
Affymetrix Human Clariom D Assay TC0100008594.hg.1 NM_001083592.1 Refseq
Affymetrix Human Clariom D Assay TC0100008594.hg.1 NM_001083592.1 Refseq
Affymetrix Human Clariom D Assay TC0100008594.hg.1 NM_005012.3 Refseq
Affymetrix Human Clariom D Assay TC0100008594.hg.1 NM_005012.3 Refseq
Affymetrix Human Clariom D Assay TC0100008594.hg.1 NM_005012.3 Refseq
Affymetrix Human Clariom D Assay TC0100008594.hg.1 NM_005012.3 Refseq
Affymetrix Human Clariom D Assay TC0100008594.hg.1 NM_005012.3 Refseq
Affymetrix Human Clariom D Assay TC0100008594.hg.1 NM_005012.3 Refseq
Affymetrix Human Clariom D Assay TC0100008594.hg.1 NM_005012.3 Refseq
Affymetrix Human Clariom D Assay TC0100008594.hg.1 NM_005012.3 Refseq
Affymetrix Human Clariom D Assay TC0100008594.hg.1 NM_005012.3 Refseq
Affymetrix Human Clariom D Assay TC0100008594.hg.1 NM_005012.3 Refseq
Affymetrix Human Clariom D Assay TC0100008594.hg.1 NM_005012.3 Refseq
Affymetrix Human Clariom D Assay TC0100008594.hg.1 NM_005012.3 Refseq
Affymetrix Human Clariom D Assay TC0100008594.hg.1 NM_005012.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0