This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05009627_s1
See other PROX2 GE Assays ›
Gene Symbol:
PROX2
Gene Name:
prospero homeobox 2
Gene Aliases:
PROX-2
Chromosome Location:
Chr.14: 74853033 - 74876663 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05009627_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PROX2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080408.2 NP_001073877.2 3 3048 95
NM_001243007.1 NP_001229936.1 4 3729 95
GenBank mRNA AK094068.1 - 4 2709 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

homeodomain transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
neuron differentiation
positive regulation of transcription from RNA polymerase II promoter
cardiac muscle cell differentiation
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 ENST00000556084.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 ENST00000556084.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 ENST00000556084.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 ENST00000556084.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 ENST00000556084.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 ENST00000556084.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 ENST00000556489.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 ENST00000556489.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 ENST00000556489.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 ENST00000556489.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 ENST00000556489.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 ENST00000556489.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 ENST00000556489.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 ENST00000619947.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 ENST00000619947.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 ENST00000619947.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 ENST00000619947.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 NM_001080408.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 NM_001080408.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 NM_001080408.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 NM_001080408.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 NM_001080408.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 NM_001243007.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 NM_001243007.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 NM_001243007.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 NM_001243007.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 NM_001243007.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 NM_001243007.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009702.hg.1 NM_001243007.1 Refseq
Affymetrix Human Clariom D Assay TC1400009702.hg.1 ENST00000556084.6 Ensembl
Affymetrix Human Clariom D Assay TC1400009702.hg.1 ENST00000556084.6 Ensembl
Affymetrix Human Clariom D Assay TC1400009702.hg.1 ENST00000556084.6 Ensembl
Affymetrix Human Clariom D Assay TC1400009702.hg.1 ENST00000556084.6 Ensembl
Affymetrix Human Clariom D Assay TC1400009702.hg.1 ENST00000556084.6 Ensembl
Affymetrix Human Clariom D Assay TC1400009702.hg.1 ENST00000556084.6 Ensembl
Affymetrix Human Clariom D Assay TC1400009702.hg.1 ENST00000556489.3 Ensembl
Affymetrix Human Clariom D Assay TC1400009702.hg.1 ENST00000556489.3 Ensembl
Affymetrix Human Clariom D Assay TC1400009702.hg.1 ENST00000556489.3 Ensembl
Affymetrix Human Clariom D Assay TC1400009702.hg.1 ENST00000556489.3 Ensembl
Affymetrix Human Clariom D Assay TC1400009702.hg.1 ENST00000556489.3 Ensembl
Affymetrix Human Clariom D Assay TC1400009702.hg.1 ENST00000556489.3 Ensembl
Affymetrix Human Clariom D Assay TC1400009702.hg.1 ENST00000556489.3 Ensembl
Affymetrix Human Clariom D Assay TC1400009702.hg.1 ENST00000619947.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009702.hg.1 ENST00000619947.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009702.hg.1 ENST00000619947.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009702.hg.1 ENST00000619947.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009702.hg.1 NM_001080408.2 Refseq
Affymetrix Human Clariom D Assay TC1400009702.hg.1 NM_001080408.2 Refseq
Affymetrix Human Clariom D Assay TC1400009702.hg.1 NM_001080408.2 Refseq
Affymetrix Human Clariom D Assay TC1400009702.hg.1 NM_001080408.2 Refseq
Affymetrix Human Clariom D Assay TC1400009702.hg.1 NM_001080408.2 Refseq
Affymetrix Human Clariom D Assay TC1400009702.hg.1 NM_001243007.1 Refseq
Affymetrix Human Clariom D Assay TC1400009702.hg.1 NM_001243007.1 Refseq
Affymetrix Human Clariom D Assay TC1400009702.hg.1 NM_001243007.1 Refseq
Affymetrix Human Clariom D Assay TC1400009702.hg.1 NM_001243007.1 Refseq
Affymetrix Human Clariom D Assay TC1400009702.hg.1 NM_001243007.1 Refseq
Affymetrix Human Clariom D Assay TC1400009702.hg.1 NM_001243007.1 Refseq
Affymetrix Human Clariom D Assay TC1400009702.hg.1 NM_001243007.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0