This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05008816_m1
See other FBLN5 GE Assays ›
Gene Symbol:
FBLN5
Gene Name:
fibulin 5
Gene Aliases:
ADCL2, ARCL1A, ARMD3, DANCE, EVEC, FIBL-5, HNARMD, UP50
Chromosome Location:
Chr.14: 91869411 - 91947702 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05008816_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FBLN5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011536356.1 XP_011534658.1 11 - 12 1675 73
XM_011536357.1 XP_011534659.1 11 - 12 1830 73
XM_011536358.1 XP_011534660.1 12 - 13 1930 73
GenBank mRNA AK125600.1 - 1 - 2 1323 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

extracellular matrix structural protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
cell-matrix adhesion
extracellular matrix organization
protein localization to cell surface
secretion
elastic fiber assembly
regulation of removal of superoxide radicals
integrin binding
calcium ion binding
protein binding
protein C-terminus binding
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000554121.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000554121.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000554121.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000554468.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000554468.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000554468.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000554468.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000554468.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000554468.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000556961.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000556961.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000556961.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000556961.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000556961.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000557088.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000557088.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000557088.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000557088.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000557088.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000557088.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000557088.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000557088.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000557088.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000557088.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000557462.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000557462.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000557462.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000557462.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000557462.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000557462.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000557570.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000557570.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000557570.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000557570.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000557570.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000557570.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 ENST00000557570.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000267620.14 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000342058.8 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000554121.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000554121.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000554121.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000554468.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000554468.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000554468.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000554468.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000554468.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000554468.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000556154.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000556961.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000556961.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000556961.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000556961.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000556961.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000557088.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000557088.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000557088.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000557088.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000557088.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000557088.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000557088.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000557088.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000557088.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000557088.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000557462.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000557462.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000557462.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000557462.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000557462.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000557462.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000557570.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000557570.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000557570.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000557570.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000557570.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000557570.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 ENST00000557570.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Clariom D Assay TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Clariom D Assay TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Clariom D Assay TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Clariom D Assay TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Clariom D Assay TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Clariom D Assay TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Clariom D Assay TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Clariom D Assay TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Clariom D Assay TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Clariom D Assay TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Clariom D Assay TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Clariom D Assay TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Clariom D Assay TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Clariom D Assay TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Clariom D Assay TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Clariom D Assay TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Clariom D Assay TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Clariom D Assay TC1400009992.hg.1 NM_006329.3 Refseq
Affymetrix Human Clariom D Assay TC1400009992.hg.1 NM_006329.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0