This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05008389_s1
See other DLK1 GE Assays ›
Gene Symbol:
DLK1
Gene Name:
delta like non-canonical Notch ligand 1
Gene Aliases:
DLK, DLK-1, Delta1, FA1, PREF1, Pref-1, ZOG, pG2
Chromosome Location:
Chr.14: 100726865 - 100738224 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05008389_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DLK1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001317172.1 NP_001304101.1 6 109
NM_003836.6 NP_003827.3 5 109
GenBank mRNA AK289384.1 - 5 109
AK297918.1 - 4 109
AK313258.1 - 5 109
BC007741.2 - 5 4128269 109
BC013197.1 - 5 4347187 109
BC014015.2 - 5 4130556 109
BX357053.2 - 1 109
BX408554.2 - 1 109
CB857925.1 - 1 109
DQ017081.1 - 6 109
DQ126100.1 - 6 109
DQ138965.1 - 6 109
U15979.1 - 5 1214 106
U15981.1 - 6 109
X17544.1 - 5 109
Z12172.1 - 5 109

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DLK1

Panther Classification:

Molecular Function -

membrane-bound signaling molecule

Gene Ontology Categories:

Function(s) Process(es)

cell differentiation
negative regulation of Notch signaling pathway
molecular_function
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000392848.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000392848.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000392848.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000392848.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000392848.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000392848.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000392848.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000392848.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000555747.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000555747.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000555747.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000555747.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000555747.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000555747.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000555747.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000556051.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000556051.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000556051.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000556051.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 ENST00000556051.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010652.hg.1 ENST00000608876.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010652.hg.1 ENST00000608876.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000331224.10 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000392848.9 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000392848.9 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000392848.9 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000392848.9 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000392848.9 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000392848.9 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000392848.9 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000392848.9 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000555747.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000555747.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000555747.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000555747.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000555747.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000555747.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000555747.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000556051.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000556051.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000556051.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000556051.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 ENST00000556051.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Clariom D Assay TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Clariom D Assay TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Clariom D Assay TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Clariom D Assay TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Clariom D Assay TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Clariom D Assay TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Clariom D Assay TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Clariom D Assay TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Clariom D Assay TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Clariom D Assay TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Clariom D Assay TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Clariom D Assay TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Clariom D Assay TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Clariom D Assay TC1400010651.hg.1 NM_003836.6 Refseq
Affymetrix Human Clariom D Assay TC1400010651.hg.1 NM_003836.6 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0