This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05006784_s1
See other MIR17HG GE Assays ›
Gene Symbol:
MIR17HG MIR18A
Gene Name:
miR-17-92a-1 cluster host gene
microRNA 18a
Gene Aliases:
C13orf25, FGLDS2, LINC00048, MIHG1, MIRH1, MIRHG1, NCRNA00048, miR-17-92
MIR18, MIRN18, MIRN18A, hsa-mir-18, hsa-mir-18a, miR-18, miRNA18A, mir-18a
Chromosome Location:
Chr.13: 91347820 - 91354575 on Build GRCh38
Chromosome Location:
Chr.13: 91350751 - 91350821 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs05006784_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR17HG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_027350.1 2 1664 137
GenBank mRNA BC109081.2 - 2 1542 137
BC109082.2 - 3 1157 137
Gene Symbol: MIR18A
Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AB176708.1 - LIT3364
oncomir-1
2 1664 137

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000362279.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000362310.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000384829.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000384878.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000385012.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000385233.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000400282.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000400282.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000400282.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000400282.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000400282.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000400282.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000400282.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000400282.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_027349.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_027349.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_027349.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_027349.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_029487.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_029488.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_029489.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_029490.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_029492.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007706.hg.1 NR_029508.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000362279.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000362310.2 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000384829.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000384878.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000385012.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000385233.2 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000400282.2 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000400282.2 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000400282.2 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000400282.2 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000400282.2 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000400282.2 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000400282.2 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000400282.2 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000581816.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 ENST00000582141.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_027349.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_027349.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_027349.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_027349.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_027350.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_029487.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_029488.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_029489.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_029490.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_029492.1 Refseq
Affymetrix Human Clariom D Assay TC1300007706.hg.1 NR_029508.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0