This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05005831_m1
See other PCDH9 GE Assays ›
Gene Symbol:
PCDH9
Gene Name:
protocadherin 9
Gene Aliases:
-
Chromosome Location:
Chr.13: 66302834 - 67230336 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05005831_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PCDH9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001318372.1 NP_001305301.1 1 - 2 562 92
NM_001318373.1 NP_001305302.1 1 - 2 562 92
NM_001318374.1 NP_001305303.1 1 - 2 562 92
NM_020403.4 NP_065136.1 1 - 2 562 92
NM_203487.2 NP_982354.1 1 - 2 562 92
XM_005266408.3 XP_005266465.1 1 - 2 562 92
XM_011535099.2 XP_011533401.1 1 - 2 562 92
XM_017020620.1 XP_016876109.1 1 - 2 562 92
GenBank mRNA AK054661.1 - 1 - 2 562 92
BC136626.1 - 1 - 2 44 9020616 92
BC136627.1 - 1 - 2 44 9020617 92
BC144321.1 - 1 - 2 44 9052841 92
BC150296.1 - 1 - 2 671 8860363 92
BX537422.1 - 1 - 2 560 92
DA497692.1 - 1 - 2 84 92

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC136626.1, BC136627.1, BC144321.1, BC150296.1, BX537422.1, NM_001318372.1, NM_001318373.1, NM_001318374.1, NM_020403.4, NM_203487.2, XM_005266408.3, XM_011535099.2, XM_017020620.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PCDH9

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

homophilic cell adhesion via plasma membrane adhesion molecules
forebrain development
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000377861.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000377861.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000377861.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000377861.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000377861.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000377861.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000377861.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000377861.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000377861.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000377861.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000377861.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000377865.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000377865.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000377865.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000377865.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000377865.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000377865.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000377865.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000377865.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000377865.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000377865.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000377865.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000456367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000456367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000456367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000456367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000456367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000456367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000456367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000456367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000456367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000544246.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000544246.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000544246.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000544246.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000544246.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000544246.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000544246.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000544246.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000544246.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000544246.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000544246.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000544246.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000614931.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000614931.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000614931.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000614931.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000614931.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000617020.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000617020.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000617020.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 ENST00000617020.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 NM_020403.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 NM_020403.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 NM_020403.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 NM_020403.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 NM_020403.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 NM_020403.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 NM_020403.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 NM_020403.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 NM_020403.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 NM_020403.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 NM_020403.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 NM_203487.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 NM_203487.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 NM_203487.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 NM_203487.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 NM_203487.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 NM_203487.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 NM_203487.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 NM_203487.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 NM_203487.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 NM_203487.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 NM_203487.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009165.hg.1 NM_203487.2 Refseq
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000377861.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000377861.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000377861.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000377861.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000377861.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000377861.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000377861.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000377861.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000377861.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000377861.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000377861.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000377865.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000377865.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000377865.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000377865.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000377865.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000377865.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000377865.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000377865.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000377865.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000377865.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000377865.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000456367.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000456367.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000456367.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000456367.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000456367.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000456367.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000456367.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000456367.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000456367.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000544246.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000544246.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000544246.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000544246.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000544246.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000544246.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000544246.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000544246.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000544246.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000544246.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000544246.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000544246.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000614931.1 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000614931.1 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000614931.1 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000614931.1 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000614931.1 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000617020.1 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000617020.1 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000617020.1 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 ENST00000617020.1 Ensembl
Affymetrix Human Clariom D Assay TC1300009165.hg.1 NM_020403.4 Refseq
Affymetrix Human Clariom D Assay TC1300009165.hg.1 NM_020403.4 Refseq
Affymetrix Human Clariom D Assay TC1300009165.hg.1 NM_020403.4 Refseq
Affymetrix Human Clariom D Assay TC1300009165.hg.1 NM_020403.4 Refseq
Affymetrix Human Clariom D Assay TC1300009165.hg.1 NM_020403.4 Refseq
Affymetrix Human Clariom D Assay TC1300009165.hg.1 NM_020403.4 Refseq
Affymetrix Human Clariom D Assay TC1300009165.hg.1 NM_020403.4 Refseq
Affymetrix Human Clariom D Assay TC1300009165.hg.1 NM_020403.4 Refseq
Affymetrix Human Clariom D Assay TC1300009165.hg.1 NM_020403.4 Refseq
Affymetrix Human Clariom D Assay TC1300009165.hg.1 NM_020403.4 Refseq
Affymetrix Human Clariom D Assay TC1300009165.hg.1 NM_020403.4 Refseq
Affymetrix Human Clariom D Assay TC1300009165.hg.1 NM_203487.2 Refseq
Affymetrix Human Clariom D Assay TC1300009165.hg.1 NM_203487.2 Refseq
Affymetrix Human Clariom D Assay TC1300009165.hg.1 NM_203487.2 Refseq
Affymetrix Human Clariom D Assay TC1300009165.hg.1 NM_203487.2 Refseq
Affymetrix Human Clariom D Assay TC1300009165.hg.1 NM_203487.2 Refseq
Affymetrix Human Clariom D Assay TC1300009165.hg.1 NM_203487.2 Refseq
Affymetrix Human Clariom D Assay TC1300009165.hg.1 NM_203487.2 Refseq
Affymetrix Human Clariom D Assay TC1300009165.hg.1 NM_203487.2 Refseq
Affymetrix Human Clariom D Assay TC1300009165.hg.1 NM_203487.2 Refseq
Affymetrix Human Clariom D Assay TC1300009165.hg.1 NM_203487.2 Refseq
Affymetrix Human Clariom D Assay TC1300009165.hg.1 NM_203487.2 Refseq
Affymetrix Human Clariom D Assay TC1300009165.hg.1 NM_203487.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0