This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05004991_s1
See other PLPPR3 GE Assays ›
Gene Symbol:
PLPPR3
Gene Name:
phospholipid phosphatase related 3
Gene Aliases:
LPPR3, LPR3, PRG-2, PRG2
Chromosome Location:
Chr.19: 812488 - 824244 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05004991_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PLPPR3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001270366.1 NP_001257295.1 8 1889 73
NM_024888.2 NP_079164.1 7 1973 73
XM_011528317.2 XP_011526619.1 7 2565 73
GenBank mRNA AF357888.1 - 7 2236 73
AF541282.1 - 7 1973 73
AK021597.1 - 4 1090 73
AL136596.1 - 7 1973 73
AY304517.1 - 7 1973 73
AY436785.1 - 8 1888 73

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PLPPR3

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

phospholipid metabolic process
signal transduction
phospholipid dephosphorylation
phosphatidate phosphatase activity
lipid phosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000517665.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000517665.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000517665.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000517665.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000519502.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000519502.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000519502.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000519502.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000519502.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000519502.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000359894.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000517665.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000517665.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000517665.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000517665.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000519502.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000519502.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000519502.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000519502.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000519502.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000519502.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 ENST00000520876.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_001270366.1 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_024888.2 Refseq
Affymetrix Human Clariom D Assay TC1900009112.hg.1 NM_024888.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header