This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04996849_m1
See other PIP5K1B GE Assays ›
Gene Symbol:
PIP5K1B
Gene Name:
phosphatidylinositol-4-phosphate 5-kinase type 1 beta
Gene Aliases:
MSS4, STM7
Chromosome Location:
Chr.9: 68705217 - 69009176 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04996849_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PIP5K1B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011519083.2 XP_011517385.1 1 - 2 174 142
XM_017015189.1 XP_016870678.1 1 - 2 174 142

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011519083.2, XM_017015189.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

phosphatidylinositol biosynthetic process
regulation of phosphatidylinositol 3-kinase signaling
phosphatidylinositol phosphorylation
1-phosphatidylinositol-3-phosphate 5-kinase activity
protein binding
ATP binding
1-phosphatidylinositol-4-phosphate 5-kinase activity
1-phosphatidylinositol-5-kinase activity
1-phosphatidylinositol-3-phosphate 4-kinase activity
phosphatidylinositol-3,4-bisphosphate 5-kinase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000377284.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000377284.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000377284.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000377284.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000377284.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000377284.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000437200.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000437200.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000437200.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000437200.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000437200.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000437200.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000437200.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000440050.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000440050.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000440050.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000440050.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000440050.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000440050.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000440050.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000440050.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000472907.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000472907.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000472907.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000472907.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000472907.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000474356.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000474356.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000265382.7 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000377284.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000377284.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000377284.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000377284.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000377284.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000377284.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000437200.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000437200.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000437200.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000437200.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000437200.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000437200.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000437200.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000440050.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000440050.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000440050.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000440050.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000440050.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000440050.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000440050.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000440050.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000472907.6 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000472907.6 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000472907.6 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000472907.6 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000472907.6 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000474356.1 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000474356.1 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000478500.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 ENST00000541509.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_001278253.1 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_003558.3 Refseq
Affymetrix Human Clariom D Assay TC0900007488.hg.1 NM_003558.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0