This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04996731_g1
See other JAK2 GE Assays ›
Gene Symbol:
JAK2
Gene Name:
Janus kinase 2
Gene Aliases:
JTK10, THCYT3
Chromosome Location:
Chr.9: 4985086 - 5128183 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04996731_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: JAK2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001322194.1 NP_001309123.1 24 - 25 3885 123
NM_001322195.1 NP_001309124.1 23 - 24 3403 123
NM_001322196.1 NP_001309125.1 23 - 24 3716 123
NM_001322198.1 NP_001309127.1 24 - 25 3704 123
NM_001322199.1 NP_001309128.1 24 - 25 3646 123
NM_001322204.1 NP_001309133.1 21 - 22 3155 123
NM_004972.3 NP_004963.1 24 - 25 3784 123
GenBank mRNA AF001362.1 - 22 - 23 3290 123
AF005216.1 - 24 - 25 3784 123
AF058925.1 - 24 - 25 3767 123
AK292525.1 - 23 - 24 3403 123
AK302618.1 - 21 - 22 3214 123
AK308668.1 - 3 - 4 398 123
AY973034.1 - 23 - 24 3361 123
BC039695.1 - 23 - 24 3388 5556026 123

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene JAK2

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
activation of MAPKK activity
adaptive immune response
protein phosphorylation
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
movement of cell or subcellular component
signal transduction
enzyme linked receptor protein signaling pathway
G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
JAK-STAT cascade
tyrosine phosphorylation of STAT protein
STAT protein import into nucleus
mesoderm development
blood coagulation
positive regulation of cell proliferation
negative regulation of cell proliferation
intrinsic apoptotic signaling pathway in response to oxidative stress
negative regulation of cardiac muscle cell apoptotic process
positive regulation of cell-substrate adhesion
positive regulation of phosphatidylinositol 3-kinase signaling
cell migration
peptidyl-tyrosine phosphorylation
cytokine-mediated signaling pathway
negative regulation of cell-cell adhesion
actin filament polymerization
cell differentiation
erythrocyte differentiation
positive regulation of cell migration
axon regeneration
mineralocorticoid receptor signaling pathway
positive regulation of insulin secretion
response to lipopolysaccharide
positive regulation of phosphoprotein phosphatase activity
positive regulation of interleukin-1 beta production
positive regulation of tumor necrosis factor production
positive regulation of protein import into nucleus, translocation
response to hydroperoxide
tumor necrosis factor-mediated signaling pathway
response to tumor necrosis factor
histone H3-Y41 phosphorylation
intracellular signal transduction
interleukin-12-mediated signaling pathway
peptidyl-tyrosine autophosphorylation
regulation of cell proliferation
tyrosine phosphorylation of Stat3 protein
tyrosine phosphorylation of Stat5 protein
tyrosine phosphorylation of Stat1 protein
positive regulation of tyrosine phosphorylation of Stat3 protein
positive regulation of tyrosine phosphorylation of Stat5 protein
activation of JAK2 kinase activity
regulation of apoptotic process
positive regulation of DNA binding
negative regulation of DNA binding
negative regulation of neuron apoptotic process
positive regulation of GTPase activity
innate immune response
positive regulation of nitric oxide biosynthetic process
positive regulation of cell differentiation
negative regulation of heart contraction
response to antibiotic
protein autophosphorylation
platelet-derived growth factor receptor signaling pathway
regulation of inflammatory response
positive regulation of inflammatory response
positive regulation of peptidyl-tyrosine phosphorylation
positive regulation of cell activation
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of nitric-oxide synthase biosynthetic process
interferon-gamma-mediated signaling pathway
regulation of interferon-gamma-mediated signaling pathway
growth hormone receptor signaling pathway
JAK-STAT cascade involved in growth hormone signaling pathway
positive regulation of growth hormone receptor signaling pathway
mammary gland epithelium development
response to interleukin-12
extrinsic apoptotic signaling pathway
activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
protein kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
Ras guanyl-nucleotide exchange factor activity
receptor binding
growth hormone receptor binding
interleukin-12 receptor binding
protein binding
ATP binding
protein C-terminus binding
protein kinase binding
heme binding
type 1 angiotensin receptor binding
acetylcholine receptor binding
histone kinase activity (H3-Y41 specific)
SH2 domain binding
histone binding
phosphatidylinositol 3-kinase binding
insulin receptor substrate binding
metal ion binding
peptide hormone receptor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000442675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000442675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000476574.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000476574.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000476574.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000476574.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000487310.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000487310.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000487310.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 ENST00000487310.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000381652.3 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000442675.1 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000442675.1 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000476574.5 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000476574.5 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000476574.5 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000476574.5 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000487310.1 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000487310.1 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000487310.1 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 ENST00000487310.1 Ensembl
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq
Affymetrix Human Clariom D Assay TC0900006538.hg.1 NM_004972.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0