This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04992560_s1
See other THSD7A GE Assays ›
Gene Symbol:
THSD7A
Gene Name:
thrombospondin type 1 domain containing 7A
Gene Aliases:
-
Chromosome Location:
Chr.7: 11370435 - 11832198 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04992560_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: THSD7A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015204.2 NP_056019.1 28 10016 122
XM_006715659.1 XP_006715722.1 27 9927 122
XM_006715660.1 XP_006715723.1 26 9721 122
XM_006715662.1 XP_006715725.1 25 9633 122
XM_011515193.2 XP_011513495.1 28 12340 122
XM_011515194.1 XP_011513496.1 28 9781 122
GenBank mRNA BC030083.1 - 1 325 122

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

cell adhesion molecule

Gene Ontology Categories:

Function(s)

angiogenesis
cell differentiation

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000408005.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000408005.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000408005.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000408005.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000408005.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000408005.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000480061.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000480061.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000497575.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000497575.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000497575.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000408005.2 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000408005.2 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000408005.2 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000408005.2 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000408005.2 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000408005.2 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000423059.8 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000480061.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000480061.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000497575.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000497575.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000497575.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 ENST00000617773.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq
Affymetrix Human Clariom D Assay TC0700010281.hg.1 NM_015204.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0