This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04992151_s1
See other GNAI1 GE Assays ›
Gene Symbol:
GNAI1
Gene Name:
G protein subunit alpha i1
Gene Aliases:
Gi
Chromosome Location:
Chr.7: 80134824 - 80219409 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04992151_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GNAI1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001256414.1 NP_001243343.1 8 2573 111
NM_002069.5 NP_002060.4 8 2710 111
GenBank mRNA BC026326.1 - 8 2709 4791696 111
CD299597.1 - 182 111

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GNAI1

Panther Classification:

Molecular Function -

heterotrimeric G-protein

Gene Ontology Categories:

Function(s) Process(es)

protein folding
cell cycle
G-protein coupled receptor signaling pathway
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
response to peptide hormone
regulation of cAMP-mediated signaling
negative regulation of synaptic transmission
cell division
cellular response to forskolin
magnesium ion binding
G-protein coupled receptor binding
GTPase activity
signal transducer activity
protein binding
GTP binding
GDP binding
G-protein beta/gamma-subunit complex binding
G-protein coupled serotonin receptor binding
GTPase activating protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000418742.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000418742.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000418742.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000418742.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000418742.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000442586.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000442586.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000442586.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000442586.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000442586.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000442586.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000447650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000447650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000447650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000457358.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000457358.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000457358.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000457358.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000457358.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000457358.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000457358.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000457358.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000457358.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000490206.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000490206.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000490206.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000490206.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000490206.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 ENST00000490206.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 NM_001256414.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 NM_001256414.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 NM_001256414.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 NM_001256414.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 NM_001256414.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 NM_001256414.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 NM_001256414.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 NM_001256414.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 NM_001256414.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 NM_001256414.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 NM_001256414.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 NM_001256414.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000351004.7 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000418742.5 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000418742.5 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000418742.5 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000418742.5 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000418742.5 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000442586.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000442586.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000442586.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000442586.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000442586.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000442586.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000447650.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000447650.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000447650.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000457358.6 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000457358.6 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000457358.6 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000457358.6 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000457358.6 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000457358.6 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000457358.6 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000457358.6 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000457358.6 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000490206.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000490206.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000490206.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000490206.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000490206.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 ENST00000490206.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008181.hg.1 NM_001256414.1 Refseq
Affymetrix Human Clariom D Assay TC0700008181.hg.1 NM_001256414.1 Refseq
Affymetrix Human Clariom D Assay TC0700008181.hg.1 NM_001256414.1 Refseq
Affymetrix Human Clariom D Assay TC0700008181.hg.1 NM_001256414.1 Refseq
Affymetrix Human Clariom D Assay TC0700008181.hg.1 NM_001256414.1 Refseq
Affymetrix Human Clariom D Assay TC0700008181.hg.1 NM_001256414.1 Refseq
Affymetrix Human Clariom D Assay TC0700008181.hg.1 NM_001256414.1 Refseq
Affymetrix Human Clariom D Assay TC0700008181.hg.1 NM_001256414.1 Refseq
Affymetrix Human Clariom D Assay TC0700008181.hg.1 NM_001256414.1 Refseq
Affymetrix Human Clariom D Assay TC0700008181.hg.1 NM_001256414.1 Refseq
Affymetrix Human Clariom D Assay TC0700008181.hg.1 NM_001256414.1 Refseq
Affymetrix Human Clariom D Assay TC0700008181.hg.1 NM_001256414.1 Refseq
Affymetrix Human Clariom D Assay TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Clariom D Assay TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Clariom D Assay TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Clariom D Assay TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Clariom D Assay TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Clariom D Assay TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Clariom D Assay TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Clariom D Assay TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Clariom D Assay TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Clariom D Assay TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Clariom D Assay TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Clariom D Assay TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Clariom D Assay TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Clariom D Assay TC0700008181.hg.1 NM_002069.5 Refseq
Affymetrix Human Clariom D Assay TC0700008181.hg.1 NM_002069.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0