This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04981212_s1
See other CBL GE Assays ›
Gene Symbol:
CBL
Gene Name:
Cbl proto-oncogene
Gene Aliases:
C-CBL, CBL2, FRA11B, NSLL, RNF55
Chromosome Location:
Chr.11: 119206276 - 119308149 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04981212_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CBL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005188.3 NP_005179.2 16 2981 81
XM_017018511.1 XP_016874000.1 15 2416 81
GenBank mRNA BC132733.1 - 16 2910 40146755 81
BC136463.1 - 16 2910 9020452 81
X57110.1 - 16 2987 81
X69207.1 - 1 121 81

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CBL

Panther Classification:

Molecular Function -

ligase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
cell surface receptor signaling pathway
epidermal growth factor receptor signaling pathway
transforming growth factor beta receptor signaling pathway
fibroblast growth factor receptor signaling pathway
positive regulation of phosphatidylinositol 3-kinase signaling
protein ubiquitination
negative regulation of epidermal growth factor receptor signaling pathway
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
negative regulation of apoptotic process
positive regulation of epidermal growth factor receptor signaling pathway
positive regulation of receptor-mediated endocytosis
cell-cell adhesion
phosphotyrosine binding
transcription factor activity, sequence-specific DNA binding
ubiquitin-protein transferase activity
signal transducer activity
calcium ion binding
protein binding
zinc ion binding
ligase activity
SH3 domain binding
protein kinase binding
ephrin receptor binding
ubiquitin protein ligase activity
cadherin binding involved in cell-cell adhesion

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header