This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04970794_m1
See other ADCY5 GE Assays ›
Gene Symbol:
ADCY5
Gene Name:
adenylate cyclase 5
Gene Aliases:
AC5, FDFM
Chromosome Location:
Chr.3: 123282296 - 123448632 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04970794_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADCY5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005247077.3 XP_005247134.1 16 - 17 3015 73
XM_005247078.1 XP_005247135.1 16 - 17 1953 73
XM_006713483.1 XP_006713546.1 16 - 17 1949 73
XM_006713484.1 XP_006713547.1 16 - 17 1954 73
XM_011512359.2 XP_011510661.1 16 - 17 2813 73
XM_011512360.2 XP_011510662.1 16 - 17 2010 73
XM_011512361.1 XP_011510663.1 16 - 17 1917 73
XM_017005638.1 XP_016861127.1 16 - 17 1845 73
XM_017005639.1 XP_016861128.1 16 - 17 1845 73
XM_017005640.1 XP_016861129.1 16 - 17 1941 73
GenBank mRNA AK124691.1 - 17 - 18 2365 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

adenosine receptor signaling pathway
renal water homeostasis
cAMP biosynthetic process
adenylate cyclase-activating G-protein coupled receptor signaling pathway
activation of adenylate cyclase activity
adenylate cyclase-activating dopamine receptor signaling pathway
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
adenylate cyclase-inhibiting dopamine receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
locomotory behavior
cAMP-mediated signaling
activation of protein kinase A activity
neuromuscular process controlling balance
regulation of insulin secretion involved in cellular response to glucose stimulus
cellular response to glucagon stimulus
cellular response to forskolin
adenylate cyclase activity
ATP binding
adenylate cyclase binding
metal ion binding
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0