This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04965569_s1
See other DHRS13 GE Assays ›
Gene Symbol:
DHRS13
Gene Name:
dehydrogenase/reductase 13
Gene Aliases:
SDR7C5
Chromosome Location:
Chr.17: 28897781 - 28903071 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04965569_s1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DHRS13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144683.3 NP_653284.2 5 1746 124
GenBank mRNA AK296017.1 - 3 1397 124
AY358566.1 - 5 1746 124
BC015582.2 - 4 1736 4637504 124
BQ052759.1 - 1 566 124
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AX795353.1 - 1 Chr. 17 - 28897781 - 28898139 [+] on Build GRCh38 124

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DHRS13

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

oxidation-reduction process
oxidoreductase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700007405.hg.1 ENST00000579187.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007405.hg.1 ENST00000579187.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007405.hg.1 ENST00000579187.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007405.hg.1 ENST00000579187.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007405.hg.1 ENST00000580603.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007405.hg.1 ENST00000580603.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007405.hg.1 ENST00000580603.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007405.hg.1 ENST00000580603.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007405.hg.1 ENST00000580603.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007405.hg.1 ENST00000580603.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007405.hg.1 ENST00000580603.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007405.hg.1 ENST00000580603.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007405.hg.1 ENST00000579187.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007405.hg.1 ENST00000579187.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007405.hg.1 ENST00000579187.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007405.hg.1 ENST00000579187.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007405.hg.1 ENST00000580603.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007405.hg.1 ENST00000580603.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007405.hg.1 ENST00000580603.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007405.hg.1 ENST00000580603.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007405.hg.1 ENST00000580603.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007405.hg.1 ENST00000580603.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007405.hg.1 ENST00000580603.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007405.hg.1 ENST00000580603.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0