This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04943182_s1
See other SNORA38B GE Assays ›
Gene Symbol:
SNORA38B
Gene Name:
small nucleolar RNA, H/ACA box 38B
Gene Aliases:
-
Chromosome Location:
Chr.17: 67740670 - 67740801 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04943182_s1
Size
Availability Made To Order
Catalog # 4426961
Price 4,437.00
Your Price
Online offer:
4437.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SNORA38B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_003706.2 1 87 58

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream phosphatidylinositol transfer protein,
cytoplasmic 1
PITPNC1 M-RDGB-beta,
MRDGBbeta,
RDGB-BETA,
RDGBB,
RDGBB1
-43.4kb Sense Chr. 17 - 67377281 - 67697263 on NCBI Build GRCh38
Downstream bromodomain PHD finger transcription factor BPTF FAC1,
FALZ,
NURF301
84.7kb Sense Chr. 17 - 67825517 - 67984378 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1200012131.hg.1 ENST00000364172.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008705.hg.1 ENST00000363524.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008705.hg.1 NR_003706.2 Refseq
Affymetrix Human Clariom D Assay TC1200012131.hg.1 ENST00000364172.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008705.hg.1 ENST00000363524.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008705.hg.1 NR_003706.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0