This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04942750_s1
See other LOC101927377 GE Assays ›
Gene Symbol:
LOC101927377
Gene Name:
uncharacterized LOC101927377
Gene Aliases:
-
Chromosome Location:
Chr.20: 47352561 - 47354633 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04942750_s1
Size
Availability Made To Order
Catalog # 4426961
Price 4,437.00
Your Price
Online offer:
4437.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101927377
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_134501.1 2 1622 98
GenBank mRNA BC047609.1 - 2 1622 98

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC100131496 LOC100131496 -32.3kb Sense Chr. 20 - 47318502 - 47320754 on NCBI Build GRCh38
Downstream nuclear receptor coactivator 3 NCOA3 ACTR,
AIB-1,
AIB1,
CAGH16,
CTG26,
KAT13B,
RAC3,
SRC-3,
SRC3,
TNRC14,
TNRC16,
TRAM-1,
bHLHe42,
pCIP
147.2kb Sense Chr. 20 - 47501857 - 47656877 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2000007559.hg.1 ENST00000413818.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007559.hg.1 ENST00000413818.2 Ensembl
Affymetrix Human Clariom D Assay TC2000007559.hg.1 ENST00000413818.2 Ensembl
Affymetrix Human Clariom D Assay TC2000007559.hg.1 ENST00000413818.2 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0