This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04942243_m1
See other SCEL-AS1 GE Assays ›
Gene Symbol:
SCEL-AS1
Gene Name:
SCEL antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.13: 77599755 - 77606551 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04942243_m1
Size
Availability Made To Order
Catalog # 4426961
Price 4,437.00
Your Price
Online offer:
4437.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SCEL-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_126412.1 2 - 3 196 82
GenBank mRNA AI798835.1 - 2 - 3 82

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream MYC binding protein 2,
E3 ubiquitin protein ligase
MYCBP2 Myc-bp2,
PAM,
Phr
-275.1kb Sense Chr. 13 - 77044657 - 77327066 on NCBI Build GRCh38
Downstream putative UPF0607 protein ENSP00000383144 LOC100129307 57.0kb Sense Chr. 13 - 77661374 - 77662803 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300009303.hg.1 ENST00000456280.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009303.hg.1 ENST00000456280.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009303.hg.1 ENST00000456280.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009303.hg.1 ENST00000456280.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009303.hg.1 ENST00000457528.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009303.hg.1 ENST00000457528.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009303.hg.1 ENST00000457528.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009303.hg.1 ENST00000457528.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009303.hg.1 ENST00000457528.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009303.hg.1 ENST00000457528.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009303.hg.1 ENST00000457528.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009303.hg.1 ENST00000457528.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009303.hg.1 ENST00000457528.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009303.hg.1 ENST00000457528.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009303.hg.1 NR_126412.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009303.hg.1 NR_126412.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009303.hg.1 NR_126412.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009303.hg.1 NR_126412.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009303.hg.1 NR_126412.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009303.hg.1 NR_126412.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009303.hg.1 NR_126413.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009303.hg.1 NR_126413.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009303.hg.1 NR_126413.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009303.hg.1 NR_126413.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009303.hg.1 NR_126413.1 Refseq
Affymetrix Human Clariom D Assay TC1300009303.hg.1 ENST00000456280.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009303.hg.1 ENST00000456280.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009303.hg.1 ENST00000456280.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009303.hg.1 ENST00000456280.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009303.hg.1 ENST00000457528.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009303.hg.1 ENST00000457528.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009303.hg.1 ENST00000457528.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009303.hg.1 ENST00000457528.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009303.hg.1 ENST00000457528.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009303.hg.1 ENST00000457528.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009303.hg.1 ENST00000457528.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009303.hg.1 ENST00000457528.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009303.hg.1 ENST00000457528.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009303.hg.1 ENST00000457528.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009303.hg.1 NR_126412.1 Refseq
Affymetrix Human Clariom D Assay TC1300009303.hg.1 NR_126412.1 Refseq
Affymetrix Human Clariom D Assay TC1300009303.hg.1 NR_126412.1 Refseq
Affymetrix Human Clariom D Assay TC1300009303.hg.1 NR_126412.1 Refseq
Affymetrix Human Clariom D Assay TC1300009303.hg.1 NR_126412.1 Refseq
Affymetrix Human Clariom D Assay TC1300009303.hg.1 NR_126412.1 Refseq
Affymetrix Human Clariom D Assay TC1300009303.hg.1 NR_126413.1 Refseq
Affymetrix Human Clariom D Assay TC1300009303.hg.1 NR_126413.1 Refseq
Affymetrix Human Clariom D Assay TC1300009303.hg.1 NR_126413.1 Refseq
Affymetrix Human Clariom D Assay TC1300009303.hg.1 NR_126413.1 Refseq
Affymetrix Human Clariom D Assay TC1300009303.hg.1 NR_126413.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0