This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04942229_m1
See other MIR217HG GE Assays ›
Gene Symbol:
MIR217HG
Gene Name:
MIR217 host gene
Gene Aliases:
MIR216AHG, MIR216BHG
Chromosome Location:
Chr.2: 55963406 - 56047326 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04942229_m1
Size
Availability Made To Order
Catalog # 4426961
Price 4,437.00
Your Price
Online offer:
4437.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR217HG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_126406.1 2 - 3 208 105
GenBank mRNA DA732292.1 - 2 - 3 208 105

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC105374690 LOC105374690 Anti-sense Chr. 2 - 55945824 - 56177554 on NCBI Build GRCh38
Downstream uncharacterized LOC101927165 LOC101927165 60.9kb Sense Chr. 2 - 56074654 - 56090382 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200012660.hg.1 ENST00000446139.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012660.hg.1 ENST00000446139.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012660.hg.1 ENST00000446139.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012660.hg.1 ENST00000446139.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012660.hg.1 NR_126406.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012660.hg.1 NR_126406.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012660.hg.1 NR_126406.1 Refseq
Affymetrix Human Clariom D Assay TC0200012660.hg.1 ENST00000446139.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012660.hg.1 ENST00000446139.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012660.hg.1 ENST00000446139.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012660.hg.1 ENST00000446139.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012660.hg.1 NR_126406.1 Refseq
Affymetrix Human Clariom D Assay TC0200012660.hg.1 NR_126406.1 Refseq
Affymetrix Human Clariom D Assay TC0200012660.hg.1 NR_126406.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0