This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04942215_m1
See other LINC01219 GE Assays ›
Gene Symbol:
LINC01219
Gene Name:
long intergenic non-protein coding RNA 1219
Gene Aliases:
-
Chromosome Location:
Chr.11: 1991096 - 1993469 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04942215_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LINC01219
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_126400.1 1 - 2 169 78
GenBank mRNA BX092271.1 - 1 - 2 170 78
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CR740877.1 - 1 - 2 169 Chr. 11 - 1991097 - 1993462 [+] on Build GRCh38 78
GenBankmRNA DB258193.1 - 1 - 2 267 Chr. 11|NT_187376.1 - 46174 - 47413 [+] on Build GRCh38 78

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream MRPL23 antisense RNA 1 MRPL23-AS1 -1.2kb Anti-sense Chr. 11 - 1983209 - 1989920 on NCBI Build GRCh38
Downstream H19 opposite tumor suppressor HOTS 128.0 Sense Chr. 11 - 1992408 - 1996294 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100006549.hg.1 ENST00000418612.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006549.hg.1 ENST00000418612.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006549.hg.1 ENST00000418612.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006549.hg.1 NM_001293171.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100006549.hg.1 NM_001293171.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100006549.hg.1 NM_001293171.2 Refseq
Affymetrix Human Clariom D Assay TC1100006549.hg.1 ENST00000418612.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006549.hg.1 ENST00000418612.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006549.hg.1 ENST00000418612.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006549.hg.1 NM_001293171.2 Refseq
Affymetrix Human Clariom D Assay TC1100006549.hg.1 NM_001293171.2 Refseq
Affymetrix Human Clariom D Assay TC1100006549.hg.1 NM_001293171.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0