This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04942162_g1
See other LINC01537 GE Assays ›
Gene Symbol:
LINC01537
Gene Name:
long intergenic non-protein coding RNA 1537
Gene Aliases:
-
Chromosome Location:
Chr.11: 72570656 - 72573222 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04942162_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LINC01537
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_126364.1 1 - 2 251 72
GenBank mRNA AL832797.1 - 1 - 2 247 72
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BI818420.1 - 1 - 2 251 Chr. 11 - 72570656 - 72571482 [+] on Build GRCh38 72

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ClpB homolog,
mitochondrial AAA ATPase chaperonin
CLPB ANKCLB,
HSP78,
MEGCANN,
MGCA7,
SKD3
-136.0kb Anti-sense Chr. 11 - 72292425 - 72434684 on NCBI Build GRCh38
Downstream phosphodiesterase 2A PDE2A CGS-PDE,
PDE2A1,
PED2A4,
cGSPDE
2.9kb Anti-sense Chr. 11 - 72576140 - 72674453 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100008354.hg.1 ENST00000450804.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008354.hg.1 ENST00000450804.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008354.hg.1 NR_126364.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008354.hg.1 NR_126364.1 Refseq
Affymetrix Human Clariom D Assay TC1100008354.hg.1 ENST00000450804.3 Ensembl
Affymetrix Human Clariom D Assay TC1100008354.hg.1 ENST00000450804.3 Ensembl
Affymetrix Human Clariom D Assay TC1100008354.hg.1 NR_126364.1 Refseq
Affymetrix Human Clariom D Assay TC1100008354.hg.1 NR_126364.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0