This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04939655_m1
See other TNK2-AS1 GE Assays ›
Gene Symbol:
TNK2-AS1
Gene Name:
TNK2 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
-
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04939655_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TNK2-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK127609.1 - 1 - 2 699 100
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BM772281.1 - 1 - 2 473 Chr. 3 - 195908302 - 195910051 [+] on Build GRCh38 100

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 6829 MIR6829 hsa-mir-6829 -25.7kb Anti-sense Chr. 3 - 195882329 - 195882395 on NCBI Build GRCh38
Downstream succinate dehydrogenase complex flavoprotein subunit A pseudogene 1 SDHAP1 SDHAL1,
SDHALP1
48.7kb Anti-sense Chr. 3 - 195959921 - 195990279 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300009984.hg.1 ENST00000458180.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009984.hg.1 ENST00000458180.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009984.hg.1 ENST00000458180.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009984.hg.1 ENST00000458180.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0