This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04939572_g1
See other LOC150596 GE Assays ›
Gene Symbol:
LOC150596
Gene Name:
uncharacterized LOC150596
Gene Aliases:
-
Chromosome Location:
Chr.2: 149803255 - 149862438 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04939572_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC150596
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK057517.1 - 1 - 2 143 130
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB026077.1 - 1 - 2 143 Chr. 2 - 149858196 - 149859191 [-] on Build GRCh38 130

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC101929231 LOC101929231 -9.2kb Anti-sense Chr. 2 - 149587358 - 149848234 on NCBI Build GRCh38
Downstream Rho family GTPase 3 RND3 ARHE,
Rho8,
RhoE,
memB
609.0kb Sense Chr. 2 - 150468193 - 150487695 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK057517.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200014587.hg.1 ENST00000295052.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014587.hg.1 ENST00000295052.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014587.hg.1 ENST00000295052.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014587.hg.1 ENST00000295052.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014587.hg.1 ENST00000295052.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014587.hg.1 ENST00000295052.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014587.hg.1 ENST00000433174.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014587.hg.1 ENST00000433174.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014587.hg.1 ENST00000295052.3 Ensembl
Affymetrix Human Clariom D Assay TC0200014587.hg.1 ENST00000295052.3 Ensembl
Affymetrix Human Clariom D Assay TC0200014587.hg.1 ENST00000295052.3 Ensembl
Affymetrix Human Clariom D Assay TC0200014587.hg.1 ENST00000295052.3 Ensembl
Affymetrix Human Clariom D Assay TC0200014587.hg.1 ENST00000295052.3 Ensembl
Affymetrix Human Clariom D Assay TC0200014587.hg.1 ENST00000295052.3 Ensembl
Affymetrix Human Clariom D Assay TC0200014587.hg.1 ENST00000433174.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014587.hg.1 ENST00000433174.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0