This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04938895_g1
See other LOC105377716 GE Assays ›
Gene Symbol:
LOC105377716
Gene Name:
uncharacterized LOC105377716
Gene Aliases:
-
Chromosome Location:
Chr.5: 170639158 - 170681470 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04938895_g1
Size
Availability Made To Order
Catalog # 4426961
Price 4,437.00
Your Price
Online offer:
4437.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC105377716
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_136214.1 3 - 4 966 75
GenBank mRNA BX116959.1 - 2 - 3 388 75

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC101928033 CTD-2270F17.1 -255.6kb Anti-sense Chr. 5 - 170389493 - 170422844 on NCBI Build GRCh38
Downstream gamma-aminobutyric acid type A receptor pi subunit GABRP 104.5kb Anti-sense Chr. 5 - 170783719 - 170814047 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500012793.hg.1 ENST00000523591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012793.hg.1 ENST00000523591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012793.hg.1 ENST00000523591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012793.hg.1 ENST00000523591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012793.hg.1 ENST00000523591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012793.hg.1 ENST00000523591.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012793.hg.1 ENST00000523591.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012793.hg.1 ENST00000523591.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012793.hg.1 ENST00000523591.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012793.hg.1 ENST00000523591.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012793.hg.1 ENST00000523591.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012793.hg.1 ENST00000523591.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0