This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04938589_g1
See other GPR50-AS1 GE Assays ›
Gene Symbol:
GPR50-AS1
Gene Name:
GPR50 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.X: 151175198 - 151177836 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04938589_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR50-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_135300.1 2 - 3 545 120
GenBank mRNA BX343514.2 - 2 - 3 546 120
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX324423.2 - 2 - 3 544 Chr. X - 151175905 - 151177836 [-] on Build GRCh38 120

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 4330 MIR4330 -6.9kb Anti-sense Chr. X - 151168222 - 151168326 on NCBI Build GRCh38
Downstream VMA21 vacuolar H+-ATPase homolog (S. cerevisiae) VMA21 MEAX,
XMEA
219.7kb Anti-sense Chr. X - 151396555 - 151409364 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0X00011108.hg.1 ENST00000454196.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00011108.hg.1 ENST00000454196.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00011108.hg.1 ENST00000454196.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00011108.hg.1 ENST00000454196.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00011108.hg.1 ENST00000454196.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00011108.hg.1 ENST00000454196.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0