This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04938324_m1
See other LOC339978 GE Assays ›
Gene Symbol:
LOC339978
Gene Name:
uncharacterized LOC339978
Gene Aliases:
-
Chromosome Location:
Chr.4: 54603211 - 54607131 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04938324_m1
Size
Availability Made To Order
Catalog # 4426961
Price 4,437.00
Your Price
Online offer:
4437.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC339978
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_134657.1 1 - 2 249 117
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BI833813.1 - 1 - 2 250 Chr. 4 - 54604673 - 54607131 [-] on Build GRCh38 117

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream platelet derived growth factor receptor alpha PDGFRA CD140A,
GAS9,
PDGFR-2,
PDGFR2,
RHEPDGFRA
-305.0kb Anti-sense Chr. 4 - 54229097 - 54298245 on NCBI Build GRCh38
Downstream KIT proto-oncogene receptor tyrosine kinase KIT C-Kit,
CD117,
PBT,
SCFR
50.8kb Anti-sense Chr. 4 - 54657928 - 54740715 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0400010725.hg.1 ENST00000508484.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010725.hg.1 ENST00000508484.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010725.hg.1 ENST00000508484.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010725.hg.1 ENST00000508484.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0