This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04937767_m1
See other BCAR4 GE Assays ›
Gene Symbol:
BCAR4
Gene Name:
breast cancer anti-estrogen resistance 4 (non-protein coding)
Gene Aliases:
-
Chromosome Location:
Chr.16: 11819830 - 11828832 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04937767_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BCAR4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_131222.1 1 - 2 128 124
GenBank mRNA R78073.1 - 1 - 2 107 124

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream zinc finger CCCH-type containing 7A ZC3H7A HSPC055,
ZC3H7,
ZC3HDC7
-24.4kb Sense Chr. 16 - 11750586 - 11797258 on NCBI Build GRCh38
Downstream ribosomal L1 domain containing 1 RSL1D1 CSIG,
L12,
PBK1,
UTP30
5.4kb Sense Chr. 16 - 11834196 - 11851585 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000571158.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000571158.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000571158.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000571158.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000571158.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000571158.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000571158.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000571158.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000571158.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000571158.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000571158.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000571158.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000571158.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000571158.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000571259.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000571259.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000571259.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000571259.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000571259.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000571259.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000571259.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000571259.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000571259.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000571259.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000571259.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000571259.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000573037.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000573037.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000573037.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000573037.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000573037.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000573037.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000573037.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000573319.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000573319.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000573319.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000573319.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000573319.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000573319.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000573319.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000573319.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000573319.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000573319.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000574028.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000574028.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000574028.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000577041.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000577041.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000577041.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000615574.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000615574.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 ENST00000615574.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_024049.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_024049.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_024049.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_024049.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_024049.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_024049.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_024049.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_024049.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_024049.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_024049.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_024050.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_024050.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_024050.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_024050.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_024050.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_024050.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_131216.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_131216.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_131216.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_131216.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_131216.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_131217.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_131217.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_131217.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_131217.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_131217.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_131217.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_131217.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_131222.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_131222.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_131222.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_131222.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_131222.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009419.hg.1 NR_131222.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0