This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04937650_m1
See other EIF2B5-AS1 GE Assays ›
Gene Symbol:
EIF2B5-AS1
Gene Name:
EIF2B5 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 184546714 - 184556918 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04937650_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF2B5-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_046593.1 1 - 2 105 123
GenBank mRNA DA918749.1 - 1 - 2 105 123

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC105374250 LOC105374250 -95.6kb Anti-sense Chr. 3 - 184401537 - 184451071 on NCBI Build GRCh38
Downstream EPH receptor B3 EPHB3 EK2,
ETK2,
HEK2,
TYRO6
4.9kb Anti-sense Chr. 3 - 184561799 - 184582408 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300013393.hg.1 ENST00000421870.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013393.hg.1 ENST00000421870.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013393.hg.1 NR_046593.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013393.hg.1 NR_046593.1 Refseq
Affymetrix Human Clariom D Assay TC0300013393.hg.1 ENST00000421870.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013393.hg.1 ENST00000421870.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013393.hg.1 NR_046593.1 Refseq
Affymetrix Human Clariom D Assay TC0300013393.hg.1 NR_046593.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0