Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009607.hg.1 |
ENST00000545526.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009607.hg.1 |
ENST00000545526.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009607.hg.1 |
NR_046579.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009607.hg.1 |
NR_046579.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200171834.hg.1 |
ENST00000545526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171834.hg.1 |
NR_046579.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200171834.hg.1 |
ENST00000545526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171834.hg.1 |
NR_046579.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200171834.hg.1 |
TC1200009607.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200171835.hg.1 |
ENST00000545526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171835.hg.1 |
NR_046579.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200171835.hg.1 |
ENST00000545526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171835.hg.1 |
NR_046579.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200171835.hg.1 |
TC1200009607.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200009607.hg.1 |
ENST00000545526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009607.hg.1 |
ENST00000545526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009607.hg.1 |
NR_046579.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009607.hg.1 |
NR_046579.1 |
Refseq |