This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04415488_s1
See other LOC644762 GE Assays ›
Gene Symbol:
LOC644762
Gene Name:
mitochondrial fission factor pseudogene
Gene Aliases:
-
Chromosome Location:
Chr.5: 149931358 - 149932977 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04415488_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC644762
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110534.1 1 153 109

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream PPARG coactivator 1 beta PPARGC1B ERRL1,
PERC,
PGC-1(beta),
PGC1B
-73.5kb Anti-sense Chr. 5 - 149730252 - 149857861 on NCBI Build GRCh38
Downstream solute carrier family 26 member 2 SLC26A2 D5S1708,
DTD,
DTDST,
EDM4,
MST153,
MSTP157
27.8kb Anti-sense Chr. 5 - 149960729 - 149987400 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500012446.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500012446.hg.1 ENST00000503102.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012446.hg.1 NR_110534.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012446.hg.1 NR_110534.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012446.hg.1 NR_110534.1 Refseq
Affymetrix Human Clariom D Assay PSR0500186643.hg.1 ENST00000503102.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500186643.hg.1 NR_110534.1 Refseq
Affymetrix Human Clariom D Assay PSR0500186643.hg.1 ENST00000503102.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500186643.hg.1 NR_110534.1 Refseq
Affymetrix Human Clariom D Assay PSR0500186643.hg.1 TC0500012446.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500186644.hg.1 NR_110534.1 Refseq
Affymetrix Human Clariom D Assay PSR0500186644.hg.1 NR_110534.1 Refseq
Affymetrix Human Clariom D Assay PSR0500186644.hg.1 TC0500012446.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0500012446.hg.1 ENST00000503102.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012446.hg.1 NR_110534.1 Refseq
Affymetrix Human Clariom D Assay TC0500012446.hg.1 NR_110534.1 Refseq
Affymetrix Human Clariom D Assay TC0500012446.hg.1 NR_110534.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0