This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04409654_s1
See other FLG-AS1 GE Assays ›
Gene Symbol:
FLG-AS1
Gene Name:
FLG antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 152313459 - 152366692 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04409654_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FLG-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_103778.1 7 3571 91
NR_103779.1 3 2316 91
GenBank mRNA BC041408.1 - 3 2320 91
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX107494.1 - 1 370 Chr. 1 - 152365782 - 152366581 [+] on Build GRCh38 91
GenBankmRNA CB046878.1 - 1 198 Chr. 1 - 152365977 - 152366462 [+] on Build GRCh38 91

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream filaggrin family member 2 FLG2 IFPS -4.0kb Anti-sense Chr. 1 - 152348735 - 152360028 on NCBI Build GRCh38
Downstream cornulin CRNN C1orf10,
DRC1,
PDRC1,
SEP53
42.6kb Anti-sense Chr. 1 - 152409243 - 152414274 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100018291.hg.1

Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000629331.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000629331.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103779.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103779.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103779.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103779.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103779.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103779.1 Refseq
Affymetrix Human Clariom D Assay PSR0100239430.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100239430.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100239430.hg.1 NR_103779.1 Refseq
Affymetrix Human Clariom D Assay PSR0100239430.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100239430.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100239430.hg.1 NR_103779.1 Refseq
Affymetrix Human Clariom D Assay PSR0100239430.hg.1 TC0100018291.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000629331.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000629331.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103779.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103779.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103779.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103779.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103779.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103779.1 Refseq
Affymetrix Human Clariom D Assay PSR0100239436.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100239436.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100239436.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay PSR0100239436.hg.1 NR_103779.1 Refseq
Affymetrix Human Clariom D Assay PSR0100239436.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100239436.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100239436.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay PSR0100239436.hg.1 NR_103779.1 Refseq
Affymetrix Human Clariom D Assay PSR0100239436.hg.1 TC0100018291.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100239437.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100239437.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100239437.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay PSR0100239437.hg.1 NR_103779.1 Refseq
Affymetrix Human Clariom D Assay PSR0100239437.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100239437.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100239437.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay PSR0100239437.hg.1 NR_103779.1 Refseq
Affymetrix Human Clariom D Assay PSR0100239437.hg.1 TC0100018291.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 234922 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0