Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013562.hg.1 |
ENST00000415477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013562.hg.1 |
ENST00000415477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013562.hg.1 |
ENST00000415477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013562.hg.1 |
ENST00000415477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013562.hg.1 |
ENST00000415477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013562.hg.1 |
ENST00000431947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013562.hg.1 |
ENST00000431947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013562.hg.1 |
NR_125868.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013562.hg.1 |
NR_125868.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013562.hg.1 |
NR_125868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013562.hg.1 |
ENST00000415477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013562.hg.1 |
ENST00000415477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013562.hg.1 |
ENST00000415477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013562.hg.1 |
ENST00000415477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013562.hg.1 |
ENST00000415477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013562.hg.1 |
ENST00000431947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013562.hg.1 |
ENST00000431947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013562.hg.1 |
NR_125868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013562.hg.1 |
NR_125868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013562.hg.1 |
NR_125868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193634.hg.1 |
ENST00000431947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193634.hg.1 |
NR_125868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193634.hg.1 |
ENST00000431947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193634.hg.1 |
NR_125868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193634.hg.1 |
TC0600013562.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600193637.hg.1 |
ENST00000415477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193637.hg.1 |
NR_125868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193637.hg.1 |
ENST00000415477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193637.hg.1 |
NR_125868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193637.hg.1 |
TC0600013562.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600193635.hg.1 |
ENST00000431947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193635.hg.1 |
NR_125868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193635.hg.1 |
ENST00000431947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193635.hg.1 |
NR_125868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193635.hg.1 |
TC0600013562.hg.1 |
Clariom D TC |