This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04408979_s1
See other LOC101926908 GE Assays ›
Gene Symbol:
LOC101926908
Gene Name:
uncharacterized LOC101926908
Gene Aliases:
-
Chromosome Location:
Chr.8: 72732045 - 72751843 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04408979_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101926908
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110656.1 3 1021 119

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream transient receptor potential cation channel subfamily A member 1 pseudogene LOC392232 -498.6kb Anti-sense Chr. 8 - 72202752 - 72251634 on NCBI Build GRCh38
Downstream telomeric repeat binding factor 1 TERF1 PIN2,
TRBF1,
TRF,
TRF1,
hTRF1-AS,
t-TRF1
257.0kb Sense Chr. 8 - 73008862 - 73047752 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0