This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04408186_m1
See other FLG-AS1 GE Assays ›
Gene Symbol:
FLG-AS1
Gene Name:
FLG antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 152313459 - 152366692 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04408186_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FLG-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_103778.1 4 - 5 1184 99
GenBank mRNA AK056431.1 - 4 - 5 1184 99

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000629331.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 ENST00000629331.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103779.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103779.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103779.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103779.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103779.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018291.hg.1 NR_103779.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000392688.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000445097.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000629331.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 ENST00000629331.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103778.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103779.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103779.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103779.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103779.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103779.1 Refseq
Affymetrix Human Clariom D Assay TC0100018291.hg.1 NR_103779.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0