Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000413192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000413192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000413192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000413192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172374.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172374.hg.1 |
NR_103561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172374.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172374.hg.1 |
NR_103561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172374.hg.1 |
TC0100009980.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100172376.hg.1 |
ENST00000413192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172376.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172376.hg.1 |
NR_103561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172376.hg.1 |
ENST00000413192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172376.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172376.hg.1 |
NR_103561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172376.hg.1 |
TC0100009980.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000413192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000413192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000413192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000413192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172375.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172375.hg.1 |
NR_103561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172375.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172375.hg.1 |
NR_103561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172375.hg.1 |
TC0100009980.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100172383.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172383.hg.1 |
NR_103561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172383.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172383.hg.1 |
NR_103561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172383.hg.1 |
TC0100009980.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100172381.hg.1 |
ENST00000413192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172381.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172381.hg.1 |
NR_103561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172381.hg.1 |
ENST00000413192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172381.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172381.hg.1 |
NR_103561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172381.hg.1 |
TC0100009980.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100172378.hg.1 |
ENST00000413192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172378.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172378.hg.1 |
NR_103561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172378.hg.1 |
ENST00000413192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172378.hg.1 |
ENST00000432386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172378.hg.1 |
NR_103561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172378.hg.1 |
TC0100009980.hg.1 |
Clariom D TC |