This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04408040_m1
See other EGFLAM-AS2 GE Assays ›
Gene Symbol:
EGFLAM-AS2
Gene Name:
EGFLAM antisense RNA 2
Gene Aliases:
-
Chromosome Location:
Chr.5: 38399814 - 38403471 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04408040_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EGFLAM-AS2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_102750.1 2 - 3 331 130
GenBank mRNA AI359548.1 - 1 - 2 130
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AW594505.1 - 1 - 2 Chr. 5 - 38399814 - 38401961 [+] on Build GRCh38 130
GenBankmRNA AA757273.1 - 1 - 2 Chr. 5 - 38399817 - 38402085 [+] on Build GRCh38 130
GenBankmRNA BF510076.1 - 1 - 2 Chr. 5 - 38399814 - 38402072 [+] on Build GRCh38 130

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream EGFLAM antisense RNA 4 EGFLAM-AS4 -108.8kb Sense Chr. 5 - 38282269 - 38290986 on NCBI Build GRCh38
Downstream leukemia inhibitory factor receptor alpha LIFR CD118,
LIF-R,
SJS2,
STWS,
SWS
72.9kb Sense Chr. 5 - 38474963 - 38606290 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500010534.hg.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 ENST00000512603.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 ENST00000512603.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 ENST00000512603.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 ENST00000512603.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 ENST00000514377.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 ENST00000514377.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 ENST00000514377.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 NR_102749.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 NR_102749.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 NR_102749.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 NR_102749.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 NR_102749.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 NR_102750.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 NR_102750.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010534.hg.1 NR_102750.1 Refseq
Affymetrix Human Clariom D Assay PSR0500173403.hg.1 ENST00000512603.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500173403.hg.1 ENST00000512603.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500173403.hg.1 NR_102750.1 Refseq
Affymetrix Human Clariom D Assay PSR0500173403.hg.1 ENST00000512603.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500173403.hg.1 ENST00000512603.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500173403.hg.1 NR_102750.1 Refseq
Affymetrix Human Clariom D Assay PSR0500173403.hg.1 TC0500010534.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500173407.hg.1 ENST00000512603.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500173407.hg.1 ENST00000512603.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500173407.hg.1 ENST00000514377.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500173407.hg.1 NR_102749.1 Refseq
Affymetrix Human Clariom D Assay PSR0500173407.hg.1 NR_102750.1 Refseq
Affymetrix Human Clariom D Assay PSR0500173407.hg.1 ENST00000512603.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500173407.hg.1 ENST00000512603.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500173407.hg.1 ENST00000514377.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500173407.hg.1 NR_102749.1 Refseq
Affymetrix Human Clariom D Assay PSR0500173407.hg.1 NR_102750.1 Refseq
Affymetrix Human Clariom D Assay PSR0500173407.hg.1 TC0500010534.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0500010534.hg.1 ENST00000512603.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010534.hg.1 ENST00000512603.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010534.hg.1 ENST00000512603.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010534.hg.1 ENST00000512603.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010534.hg.1 ENST00000514377.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010534.hg.1 ENST00000514377.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010534.hg.1 ENST00000514377.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010534.hg.1 NR_102749.1 Refseq
Affymetrix Human Clariom D Assay TC0500010534.hg.1 NR_102749.1 Refseq
Affymetrix Human Clariom D Assay TC0500010534.hg.1 NR_102749.1 Refseq
Affymetrix Human Clariom D Assay TC0500010534.hg.1 NR_102749.1 Refseq
Affymetrix Human Clariom D Assay TC0500010534.hg.1 NR_102749.1 Refseq
Affymetrix Human Clariom D Assay TC0500010534.hg.1 NR_102750.1 Refseq
Affymetrix Human Clariom D Assay TC0500010534.hg.1 NR_102750.1 Refseq
Affymetrix Human Clariom D Assay TC0500010534.hg.1 NR_102750.1 Refseq
Affymetrix Human Clariom D Assay PSR0500173410.hg.1 NR_102749.1 Refseq
Affymetrix Human Clariom D Assay PSR0500173410.hg.1 NR_102750.1 Refseq
Affymetrix Human Clariom D Assay PSR0500173410.hg.1 NR_102749.1 Refseq
Affymetrix Human Clariom D Assay PSR0500173410.hg.1 NR_102750.1 Refseq
Affymetrix Human Clariom D Assay PSR0500173410.hg.1 TC0500010534.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0