This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04408032_m1
See other UCHL1-AS1 GE Assays ›
Gene Symbol:
UCHL1-AS1
Gene Name:
UCHL1 antisense RNA 1 (head to head)
Gene Aliases:
-
Chromosome Location:
Chr.4: 41220074 - 41256727 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04408032_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UCHL1-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_102709.1 1 - 2 163 75
GenBank mRNA DB169394.1 - 1 - 2 163 75

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream amyloid beta precursor protein binding family B member 2 APBB2 FE65L,
FE65L1
-4.6kb Sense Chr. 4 - 40810027 - 41216714 on NCBI Build GRCh38
Downstream ubiquitin C-terminal hydrolase L1 UCHL1 HEL-117,
HEL-S-53,
NDGOA,
PARK5,
PGP 9.5,
PGP9.5,
PGP95,
Uch-L1
154.0 Anti-sense Chr. 4 - 41256881 - 41268429 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400010532.hg.1

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0400010532.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010532.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010532.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010532.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010532.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010532.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010532.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010532.hg.1 ENST00000510073.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010532.hg.1 ENST00000510073.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010532.hg.1 ENST00000510073.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010532.hg.1 NR_102709.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010532.hg.1 NR_102709.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010532.hg.1 NR_102709.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010532.hg.1 NR_102709.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010532.hg.1 NR_102709.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010532.hg.1 NR_102709.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010532.hg.1 NR_102709.1 Refseq
Affymetrix Human Clariom D Assay PSR0400171850.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400171850.hg.1 NR_102709.1 Refseq
Affymetrix Human Clariom D Assay PSR0400171850.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400171850.hg.1 NR_102709.1 Refseq
Affymetrix Human Clariom D Assay PSR0400171850.hg.1 TC0400010532.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0400171854.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400171854.hg.1 NR_102709.1 Refseq
Affymetrix Human Clariom D Assay PSR0400171854.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400171854.hg.1 NR_102709.1 Refseq
Affymetrix Human Clariom D Assay PSR0400171854.hg.1 TC0400010532.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0400171851.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400171851.hg.1 NR_102709.1 Refseq
Affymetrix Human Clariom D Assay PSR0400171851.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400171851.hg.1 NR_102709.1 Refseq
Affymetrix Human Clariom D Assay PSR0400171851.hg.1 TC0400010532.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0400171857.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400171857.hg.1 ENST00000510073.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400171857.hg.1 NR_102709.1 Refseq
Affymetrix Human Clariom D Assay PSR0400171857.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400171857.hg.1 ENST00000510073.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400171857.hg.1 NR_102709.1 Refseq
Affymetrix Human Clariom D Assay PSR0400171857.hg.1 TC0400010532.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0400171856.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400171856.hg.1 ENST00000510073.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400171856.hg.1 NR_102709.1 Refseq
Affymetrix Human Clariom D Assay PSR0400171856.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400171856.hg.1 ENST00000510073.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400171856.hg.1 NR_102709.1 Refseq
Affymetrix Human Clariom D Assay PSR0400171856.hg.1 TC0400010532.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0400010532.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010532.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010532.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010532.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010532.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010532.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010532.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010532.hg.1 ENST00000510073.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010532.hg.1 ENST00000510073.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010532.hg.1 ENST00000510073.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010532.hg.1 NR_102709.1 Refseq
Affymetrix Human Clariom D Assay TC0400010532.hg.1 NR_102709.1 Refseq
Affymetrix Human Clariom D Assay TC0400010532.hg.1 NR_102709.1 Refseq
Affymetrix Human Clariom D Assay TC0400010532.hg.1 NR_102709.1 Refseq
Affymetrix Human Clariom D Assay TC0400010532.hg.1 NR_102709.1 Refseq
Affymetrix Human Clariom D Assay TC0400010532.hg.1 NR_102709.1 Refseq
Affymetrix Human Clariom D Assay TC0400010532.hg.1 NR_102709.1 Refseq
Affymetrix Human Clariom D Assay PSR0400171853.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400171853.hg.1 NR_102709.1 Refseq
Affymetrix Human Clariom D Assay PSR0400171853.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400171853.hg.1 NR_102709.1 Refseq
Affymetrix Human Clariom D Assay PSR0400171853.hg.1 TC0400010532.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0400171852.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400171852.hg.1 NR_102709.1 Refseq
Affymetrix Human Clariom D Assay PSR0400171852.hg.1 ENST00000507190.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400171852.hg.1 NR_102709.1 Refseq
Affymetrix Human Clariom D Assay PSR0400171852.hg.1 TC0400010532.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0