This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04407303_m1
See other LOC101927412 GE Assays ›
Gene Symbol:
LOC101927412
Gene Name:
uncharacterized LOC101927412
Gene Aliases:
-
Chromosome Location:
Chr.1: 80535755 - 80646788 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04407303_m1
Size
Availability Made To Order
Catalog # 4426961
Price 4,437.00
Your Price
Online offer:
4437.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101927412
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_125940.1 1 - 2 182 86
GenBank mRNA BE466477.1 - 3 - 4 86

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream adhesion G protein-coupled receptor L4 ADGRL4 ELTD1,
ETL,
KPG_003
-1.5mb Anti-sense Chr. 1 - 78889764 - 79006810 on NCBI Build GRCh38
Downstream adhesion G protein-coupled receptor L2 ADGRL2 CIRL2,
CL2,
LEC1,
LPHH1,
LPHN2
765.6kb Sense Chr. 1 - 81306155 - 81992738 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100008839.hg.1

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 ENST00000418041.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 ENST00000418041.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 ENST00000418041.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 ENST00000418041.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 ENST00000418041.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 ENST00000418041.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 ENST00000418041.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 ENST00000418041.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 ENST00000443104.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 ENST00000443104.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 ENST00000443104.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 ENST00000443104.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 ENST00000443104.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 ENST00000443104.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 ENST00000443104.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 ENST00000443104.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 ENST00000443104.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 ENST00000443104.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 ENST00000443565.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 ENST00000443565.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 ENST00000443565.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 ENST00000443565.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 NR_125940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 NR_125940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 NR_125940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 NR_125940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 NR_125940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 NR_125941.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 NR_125941.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 NR_125941.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 NR_125941.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 NR_125942.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 NR_125942.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 NR_125942.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008839.hg.1 NR_125942.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0