This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04406755_m1
See other LOC100996583 GE Assays ›
Gene Symbol:
LOC100996583
Gene Name:
uncharacterized LOC100996583
Gene Aliases:
-
Chromosome Location:
Chr.1: 2566535 - 2584534 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04406755_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100996583
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_121638.1 1 - 2 70 79
GenBank mRNA CB852498.1 - 3 - 4 79

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream TNF receptor superfamily member 14 TNFRSF14 ATAR,
CD270,
HVEA,
HVEM,
LIGHTR,
TR2
-913.0 Sense Chr. 1 - 2555767 - 2565622 on NCBI Build GRCh38
Downstream family with sequence similarity 213 member B FAM213B C1orf93 4.8kb Sense Chr. 1 - 2586460 - 2591469 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100018163.hg.1

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100018163.hg.1 ENST00000424215.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018163.hg.1 ENST00000424215.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018163.hg.1 ENST00000424215.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018163.hg.1 ENST00000424215.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018163.hg.1 ENST00000456687.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018163.hg.1 ENST00000456687.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018163.hg.1 ENST00000456687.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018163.hg.1 ENST00000456687.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018163.hg.1 NR_121638.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018163.hg.1 NR_121638.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018163.hg.1 NR_121638.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018163.hg.1 NR_121638.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018163.hg.1 NR_121638.1 Refseq
Affymetrix Human Clariom D Assay PSR0100236898.hg.1 ENST00000424215.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100236898.hg.1 NR_121638.1 Refseq
Affymetrix Human Clariom D Assay PSR0100236898.hg.1 ENST00000424215.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100236898.hg.1 NR_121638.1 Refseq
Affymetrix Human Clariom D Assay PSR0100236898.hg.1 TC0100018163.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100236885.hg.1 ENST00000456687.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100236885.hg.1 NR_121638.1 Refseq
Affymetrix Human Clariom D Assay PSR0100236885.hg.1 ENST00000456687.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100236885.hg.1 NR_121638.1 Refseq
Affymetrix Human Clariom D Assay PSR0100236885.hg.1 TC0100018163.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100018163.hg.1 ENST00000424215.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018163.hg.1 ENST00000424215.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018163.hg.1 ENST00000424215.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018163.hg.1 ENST00000424215.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018163.hg.1 ENST00000456687.3 Ensembl
Affymetrix Human Clariom D Assay TC0100018163.hg.1 ENST00000456687.3 Ensembl
Affymetrix Human Clariom D Assay TC0100018163.hg.1 ENST00000456687.3 Ensembl
Affymetrix Human Clariom D Assay TC0100018163.hg.1 ENST00000456687.3 Ensembl
Affymetrix Human Clariom D Assay TC0100018163.hg.1 NR_121638.1 Refseq
Affymetrix Human Clariom D Assay TC0100018163.hg.1 NR_121638.1 Refseq
Affymetrix Human Clariom D Assay TC0100018163.hg.1 NR_121638.1 Refseq
Affymetrix Human Clariom D Assay TC0100018163.hg.1 NR_121638.1 Refseq
Affymetrix Human Clariom D Assay TC0100018163.hg.1 NR_121638.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0