This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04406508_m1
See other LOC101929073 GE Assays ›
Gene Symbol:
LOC101929073
Gene Name:
uncharacterized LOC101929073
Gene Aliases:
-
Chromosome Location:
Chr.10: 25924378 - 25933710 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04406508_m1
Size
Availability Made To Order
Catalog # 4426961
Price 4,437.00
Your Price
Online offer:
4437.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101929073
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_120650.1 3 - 4 410 173

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream long intergenic non-protein coding RNA 836 LINC00836 -191.4kb Anti-sense Chr. 10 - 25651712 - 25732935 on NCBI Build GRCh38
Downstream myosin IIIA MYO3A DFNB30 7.9kb Anti-sense Chr. 10 - 25934073 - 26212536 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000010083.hg.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000010083.hg.1 ENST00000452911.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010083.hg.1 ENST00000452911.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010083.hg.1 ENST00000452911.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010083.hg.1 ENST00000452911.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010083.hg.1 ENST00000452911.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010083.hg.1 NR_120650.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010083.hg.1 NR_120650.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010083.hg.1 NR_120650.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010083.hg.1 NR_120650.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010083.hg.1 NR_120650.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010083.hg.1 NR_120650.1 Refseq
Affymetrix Human Clariom D Assay PSR1000168686.hg.1 NR_120650.1 Refseq
Affymetrix Human Clariom D Assay PSR1000168686.hg.1 NR_120650.1 Refseq
Affymetrix Human Clariom D Assay PSR1000168686.hg.1 TC1000010083.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1000168688.hg.1 ENST00000452911.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000168688.hg.1 NR_120650.1 Refseq
Affymetrix Human Clariom D Assay PSR1000168688.hg.1 ENST00000452911.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000168688.hg.1 NR_120650.1 Refseq
Affymetrix Human Clariom D Assay PSR1000168688.hg.1 TC1000010083.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000010083.hg.1 ENST00000452911.1 Ensembl
Affymetrix Human Clariom D Assay TC1000010083.hg.1 ENST00000452911.1 Ensembl
Affymetrix Human Clariom D Assay TC1000010083.hg.1 ENST00000452911.1 Ensembl
Affymetrix Human Clariom D Assay TC1000010083.hg.1 ENST00000452911.1 Ensembl
Affymetrix Human Clariom D Assay TC1000010083.hg.1 ENST00000452911.1 Ensembl
Affymetrix Human Clariom D Assay TC1000010083.hg.1 NR_120650.1 Refseq
Affymetrix Human Clariom D Assay TC1000010083.hg.1 NR_120650.1 Refseq
Affymetrix Human Clariom D Assay TC1000010083.hg.1 NR_120650.1 Refseq
Affymetrix Human Clariom D Assay TC1000010083.hg.1 NR_120650.1 Refseq
Affymetrix Human Clariom D Assay TC1000010083.hg.1 NR_120650.1 Refseq
Affymetrix Human Clariom D Assay TC1000010083.hg.1 NR_120650.1 Refseq
Affymetrix Human Clariom D Assay PSR1000168687.hg.1 ENST00000452911.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000168687.hg.1 NR_120650.1 Refseq
Affymetrix Human Clariom D Assay PSR1000168687.hg.1 ENST00000452911.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000168687.hg.1 NR_120650.1 Refseq
Affymetrix Human Clariom D Assay PSR1000168687.hg.1 TC1000010083.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0