This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04406353_m1
See other LOC101929340 GE Assays ›
Gene Symbol:
LOC101929340
Gene Name:
uncharacterized LOC101929340
Gene Aliases:
-
Chromosome Location:
Chr.11: 124759134 - 124765925 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04406353_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101929340
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_120579.1 1 - 2 41 104
GenBank mRNA CB997328.1 - 1 - 2 68 104

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream endothelial cell adhesion molecule ESAM W117m -104.0 Anti-sense Chr. 11 - 124753123 - 124762327 on NCBI Build GRCh38
Downstream Myb/SANT DNA binding domain containing 2 MSANTD2 C11orf61 567.0 Anti-sense Chr. 11 - 124766492 - 124800879 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100009400.hg.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100009400.hg.1 ENST00000504932.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009400.hg.1 ENST00000504932.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009400.hg.1 ENST00000504932.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009400.hg.1 ENST00000504932.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009400.hg.1 ENST00000532579.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009400.hg.1 ENST00000532579.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009400.hg.1 ENST00000532579.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009400.hg.1 NR_120577.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009400.hg.1 NR_120577.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009400.hg.1 NR_120577.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009400.hg.1 NR_120577.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009400.hg.1 NR_120579.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009400.hg.1 NR_120579.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009400.hg.1 NR_120579.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009400.hg.1 NR_120579.1 Refseq
Affymetrix Human Clariom D Assay TC1100009400.hg.1 ENST00000504932.2 Ensembl
Affymetrix Human Clariom D Assay TC1100009400.hg.1 ENST00000504932.2 Ensembl
Affymetrix Human Clariom D Assay TC1100009400.hg.1 ENST00000504932.2 Ensembl
Affymetrix Human Clariom D Assay TC1100009400.hg.1 ENST00000504932.2 Ensembl
Affymetrix Human Clariom D Assay TC1100009400.hg.1 ENST00000532579.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009400.hg.1 ENST00000532579.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009400.hg.1 ENST00000532579.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009400.hg.1 NR_120577.1 Refseq
Affymetrix Human Clariom D Assay TC1100009400.hg.1 NR_120577.1 Refseq
Affymetrix Human Clariom D Assay TC1100009400.hg.1 NR_120577.1 Refseq
Affymetrix Human Clariom D Assay TC1100009400.hg.1 NR_120577.1 Refseq
Affymetrix Human Clariom D Assay TC1100009400.hg.1 NR_120579.1 Refseq
Affymetrix Human Clariom D Assay TC1100009400.hg.1 NR_120579.1 Refseq
Affymetrix Human Clariom D Assay TC1100009400.hg.1 NR_120579.1 Refseq
Affymetrix Human Clariom D Assay TC1100009400.hg.1 NR_120579.1 Refseq
Affymetrix Human Clariom D Assay PSR1100170405.hg.1 ENST00000504932.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100170405.hg.1 ENST00000532579.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100170405.hg.1 NR_120577.1 Refseq
Affymetrix Human Clariom D Assay PSR1100170405.hg.1 NR_120579.1 Refseq
Affymetrix Human Clariom D Assay PSR1100170405.hg.1 ENST00000504932.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100170405.hg.1 ENST00000532579.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100170405.hg.1 NR_120577.1 Refseq
Affymetrix Human Clariom D Assay PSR1100170405.hg.1 NR_120579.1 Refseq
Affymetrix Human Clariom D Assay PSR1100170405.hg.1 TC1100009400.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100170403.hg.1 ENST00000532579.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100170403.hg.1 NR_120579.1 Refseq
Affymetrix Human Clariom D Assay PSR1100170403.hg.1 ENST00000532579.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100170403.hg.1 NR_120579.1 Refseq
Affymetrix Human Clariom D Assay PSR1100170403.hg.1 TC1100009400.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100170404.hg.1 ENST00000504932.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100170404.hg.1 ENST00000532579.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100170404.hg.1 NR_120577.1 Refseq
Affymetrix Human Clariom D Assay PSR1100170404.hg.1 NR_120579.1 Refseq
Affymetrix Human Clariom D Assay PSR1100170404.hg.1 ENST00000504932.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100170404.hg.1 ENST00000532579.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100170404.hg.1 NR_120577.1 Refseq
Affymetrix Human Clariom D Assay PSR1100170404.hg.1 NR_120579.1 Refseq
Affymetrix Human Clariom D Assay PSR1100170404.hg.1 TC1100009400.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100170406.hg.1 ENST00000504932.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100170406.hg.1 NR_120577.1 Refseq
Affymetrix Human Clariom D Assay PSR1100170406.hg.1 NR_120579.1 Refseq
Affymetrix Human Clariom D Assay PSR1100170406.hg.1 ENST00000504932.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100170406.hg.1 NR_120577.1 Refseq
Affymetrix Human Clariom D Assay PSR1100170406.hg.1 NR_120579.1 Refseq
Affymetrix Human Clariom D Assay PSR1100170406.hg.1 TC1100009400.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0