This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04405857_m1
See other TMC3-AS1 GE Assays ›
Gene Symbol:
TMC3-AS1
Gene Name:
TMC3 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.15: 81324333 - 81443076 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04405857_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMC3-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_120365.1 6 - 7 901 95
GenBank mRNA HY226614.1 - 1 - 2 95

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream transmembrane channel like 3 TMC3 -6.1kb Anti-sense Chr. 15 - 81332419 - 81374077 on NCBI Build GRCh38
Downstream mex-3 RNA binding family member B MEX3B MEX-3B,
RKHD3,
RNF195
598.7kb Anti-sense Chr. 15 - 82041778 - 82046143 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1500008062.hg.1

Set Membership:

Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR1500158005.hg.1 NR_120365.1 Refseq
Affymetrix Human Clariom D Assay PSR1500158005.hg.1 NR_120365.1 Refseq
Affymetrix Human Clariom D Assay PSR1500158005.hg.1 TC1500008062.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0