This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04405829_m1
See other RORA-AS1 GE Assays ›
Gene Symbol:
RORA-AS1
Gene Name:
RORA antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.15: 60479178 - 60630639 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04405829_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RORA-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_120340.1 4 - 5 565 91
GenBank mRNA AI028673.1 - 1 - 2 91
AW663525.1 - 1 - 2 91
BX115309.1 - 4 - 5 394 91
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI867441.1 - 1 - 2 Chr. 15 - 60488539 - 60529724 [-] on Build GRCh38 91
GenBankmRNA AI205906.1 - 1 - 2 Chr. 15 - 60528645 - 60529724 [-] on Build GRCh38 91
GenBankmRNA CD244193.1 - 4 - 5 468 Chr. 15 - 60479276 - 60529889 [+] on Build GRCh38 91

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream interactor of little elongation complex ELL subunit 2 ICE2 BRCC1,
NARG2
-49.4kb Anti-sense Chr. 15 - 60419609 - 60479160 on NCBI Build GRCh38
Downstream RORA antisense RNA 2 RORA-AS2 151.8kb Sense Chr. 15 - 60681569 - 60687249 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1500007429.hg.1

Set Membership:

Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000558235.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000558235.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000558235.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000558235.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000558235.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000558235.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000558235.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000558235.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000558235.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000558235.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559203.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559203.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559203.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559203.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559902.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559902.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559902.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559902.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559902.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559902.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559902.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559902.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559902.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000559902.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000560280.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 ENST00000560280.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120340.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120340.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120340.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120340.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120340.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120340.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120340.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120341.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120341.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120341.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120341.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120341.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120341.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120341.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120342.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120342.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120342.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120342.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120342.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120342.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120342.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007429.hg.1 NR_120342.1 Refseq
Affymetrix Human Clariom D Assay PSR1500152825.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500152825.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500152825.hg.1 NR_120340.1 Refseq
Affymetrix Human Clariom D Assay PSR1500152825.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500152825.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500152825.hg.1 NR_120340.1 Refseq
Affymetrix Human Clariom D Assay PSR1500152825.hg.1 TC1500007429.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000501579.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000558140.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000558235.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000558235.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000558235.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000558235.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000558235.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000558235.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000558235.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000558235.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000558235.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000558235.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559203.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559203.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559203.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559203.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559824.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559902.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559902.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559902.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559902.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559902.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559902.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559902.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559902.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559902.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000559902.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000560280.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 ENST00000560280.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120339.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120339.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120339.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120339.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120339.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120339.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120339.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120339.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120339.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120339.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120339.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120340.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120340.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120340.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120340.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120340.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120340.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120340.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120341.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120341.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120341.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120341.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120341.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120341.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120341.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120342.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120342.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120342.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120342.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120342.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120342.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120342.1 Refseq
Affymetrix Human Clariom D Assay TC1500007429.hg.1 NR_120342.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0