This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04405096_g1
See other LINC01255 GE Assays ›
Gene Symbol:
LINC01255
Gene Name:
long intergenic non-protein coding RNA 1255
Gene Aliases:
-
Chromosome Location:
Chr.18: 11488570 - 11506983 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04405096_g1
Size
Availability Made To Order
Catalog # 4426961
Price 4,437.00
Your Price
Online offer:
4437.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LINC01255
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110778.1 2 - 3 305 121
NR_110779.1 3 - 4 226 121
GenBank mRNA AA844721.1 - 1 - 2 121
DB338965.1 - 1 - 2 121
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI150415.1 - 1 - 2 Chr. 18 - 11490110 - 11506968 [-] on Build GRCh38 121
GenBankmRNA AI208570.1 - 1 - 2 Chr. 18 - 11490181 - 11506977 [-] on Build GRCh38 121
GenBankmRNA AI024061.1 - 1 - 2 Chr. 18 - 11505800 - 11506968 [-] on Build GRCh38 121
GenBankmRNA AA815406.1 - 1 - 2 Chr. 18 - 11490120 - 11506983 [-] on Build GRCh38 121

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream piezo type mechanosensitive ion channel component 2 PIEZO2 C18orf30,
C18orf58,
DA3,
DA5,
FAM38B,
FAM38B2,
HsT748,
HsT771,
MWKS
-357.8kb Anti-sense Chr. 18 - 10670237 - 11149561 on NCBI Build GRCh38
Downstream solute carrier family 35 member G4 SLC35G4 AMAC1L1,
SLC35G4P
102.6kb Sense Chr. 18 - 11609558 - 11610612 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1800006699.hg.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000586145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000586145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000586145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000586947.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000586947.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000586947.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000586947.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000586947.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000586947.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000589566.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000589566.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000589566.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000589566.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000591318.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000591318.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000591318.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000591318.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000591318.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000591431.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000591431.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 ENST00000591431.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 NR_110778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 NR_110778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 NR_110778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 NR_110778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 NR_110778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 NR_110779.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 NR_110779.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 NR_110779.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006699.hg.1 NR_110779.1 Refseq
Affymetrix Human Clariom D Assay PSR1800145807.hg.1 ENST00000586145.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800145807.hg.1 ENST00000586947.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800145807.hg.1 ENST00000586947.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800145807.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800145807.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800145807.hg.1 NR_110778.1 Refseq
Affymetrix Human Clariom D Assay PSR1800145807.hg.1 NR_110779.1 Refseq
Affymetrix Human Clariom D Assay PSR1800145807.hg.1 ENST00000586145.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800145807.hg.1 ENST00000586947.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800145807.hg.1 ENST00000586947.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800145807.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800145807.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800145807.hg.1 NR_110778.1 Refseq
Affymetrix Human Clariom D Assay PSR1800145807.hg.1 NR_110779.1 Refseq
Affymetrix Human Clariom D Assay PSR1800145807.hg.1 TC1800006699.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1800145780.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800145780.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800145780.hg.1 NR_110779.1 Refseq
Affymetrix Human Clariom D Assay PSR1800145780.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800145780.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800145780.hg.1 NR_110779.1 Refseq
Affymetrix Human Clariom D Assay PSR1800145780.hg.1 TC1800006699.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000586145.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000586145.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000586145.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000586947.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000586947.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000586947.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000586947.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000586947.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000586947.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000589566.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000589566.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000589566.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000589566.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000591318.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000591318.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000591318.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000591318.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000591318.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000591431.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000591431.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 ENST00000591431.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006699.hg.1 NR_110778.1 Refseq
Affymetrix Human Clariom D Assay TC1800006699.hg.1 NR_110778.1 Refseq
Affymetrix Human Clariom D Assay TC1800006699.hg.1 NR_110778.1 Refseq
Affymetrix Human Clariom D Assay TC1800006699.hg.1 NR_110778.1 Refseq
Affymetrix Human Clariom D Assay TC1800006699.hg.1 NR_110778.1 Refseq
Affymetrix Human Clariom D Assay TC1800006699.hg.1 NR_110779.1 Refseq
Affymetrix Human Clariom D Assay TC1800006699.hg.1 NR_110779.1 Refseq
Affymetrix Human Clariom D Assay TC1800006699.hg.1 NR_110779.1 Refseq
Affymetrix Human Clariom D Assay TC1800006699.hg.1 NR_110779.1 Refseq
Affymetrix Human Clariom D Assay PSR1800145781.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800145781.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800145781.hg.1 NR_110779.1 Refseq
Affymetrix Human Clariom D Assay PSR1800145781.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800145781.hg.1 ENST00000587189.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800145781.hg.1 NR_110779.1 Refseq
Affymetrix Human Clariom D Assay PSR1800145781.hg.1 TC1800006699.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0