This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04404427_m1
See other LOC101928259 GE Assays ›
Gene Symbol:
LOC101928259
Gene Name:
uncharacterized LOC101928259
Gene Aliases:
-
Chromosome Location:
Chr.X: 72688949 - 72712348 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04404427_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101928259
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110391.1 3 - 4 159 132
GenBank mRNA AI968194.1 - 2 - 3 132
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BF223081.1 - 2 - 3 Chr. X - 72688961 - 72712347 [-] on Build GRCh38 132
GenBankmRNA BE503359.1 - 2 - 3 Chr. X - 72707846 - 72712347 [-] on Build GRCh38 132
GenBankmRNA AI336845.1 - 2 - 3 Chr. X - 72697157 - 72712350 [-] on Build GRCh38 132
GenBankmRNA BE503319.1 - 2 - 3 Chr. X - 72707852 - 72712347 [-] on Build GRCh38 132
GenBankmRNA AA927658.1 - 2 - 3 Chr. X - 72707846 - 72712348 [-] on Build GRCh38 132

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream histone deacetylase 8 HDAC8 CDA07,
CDLS5,
HD8,
HDACL1,
MRXS6,
RPD3,
WTS
-134.7kb Anti-sense Chr. X - 72329516 - 72573103 on NCBI Build GRCh38
Downstream family with sequence similarity 226 member B (non-protein coding) FAM226B CXorf50B,
LINC00246B,
NCRNA00246B
67.6kb Sense Chr. X - 72777073 - 72779095 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00007596.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0X00007596.hg.1 ENST00000420998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007596.hg.1 ENST00000420998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007596.hg.1 ENST00000420998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007596.hg.1 ENST00000420998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007596.hg.1 ENST00000420998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007596.hg.1 NR_110391.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007596.hg.1 NR_110391.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007596.hg.1 NR_110391.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007596.hg.1 NR_110391.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007596.hg.1 NR_110391.1 Refseq
Affymetrix Human Clariom D Assay TC0X00007596.hg.1 ENST00000420998.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007596.hg.1 ENST00000420998.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007596.hg.1 ENST00000420998.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007596.hg.1 ENST00000420998.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007596.hg.1 ENST00000420998.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007596.hg.1 NR_110391.1 Refseq
Affymetrix Human Clariom D Assay TC0X00007596.hg.1 NR_110391.1 Refseq
Affymetrix Human Clariom D Assay TC0X00007596.hg.1 NR_110391.1 Refseq
Affymetrix Human Clariom D Assay TC0X00007596.hg.1 NR_110391.1 Refseq
Affymetrix Human Clariom D Assay TC0X00007596.hg.1 NR_110391.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00152321.hg.1 ENST00000420998.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00152321.hg.1 NR_110391.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00152321.hg.1 ENST00000420998.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00152321.hg.1 NR_110391.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00152321.hg.1 TC0X00007596.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0X00152319.hg.1 ENST00000420998.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00152319.hg.1 NR_110391.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00152319.hg.1 ENST00000420998.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00152319.hg.1 NR_110391.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00152319.hg.1 TC0X00007596.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0X00152318.hg.1 ENST00000420998.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00152318.hg.1 NR_110391.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00152318.hg.1 ENST00000420998.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00152318.hg.1 NR_110391.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00152318.hg.1 TC0X00007596.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0X00152320.hg.1 ENST00000420998.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00152320.hg.1 NR_110391.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00152320.hg.1 ENST00000420998.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00152320.hg.1 NR_110391.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00152320.hg.1 TC0X00007596.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0