This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04402570_s1
See other MIR7849 GE Assays ›
Gene Symbol:
MIR7849
Gene Name:
microRNA 7849
Gene Aliases:
hsa-mir-7849
Chromosome Location:
Chr.4: 146408583 - 146408688 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04402570_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR7849
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_107003.1 1 57 76

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream solute carrier family 10 member 7 SLC10A7 C4orf13,
P7
Anti-sense Chr. 4 - 146253981 - 146522351 on NCBI Build GRCh38
Downstream POU class 4 homeobox 2 POU4F2 BRN3.2,
BRN3B,
Brn-3b
230.2kb Sense Chr. 4 - 146638893 - 146642471 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC0400008930.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0400008930.hg.1 ENST00000619235.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008930.hg.1 NR_107003.1 Refseq
Affymetrix Human Clariom D Assay TC0400008930.hg.1 ENST00000619235.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008930.hg.1 NR_107003.1 Refseq
Affymetrix Human Clariom D Assay PSR0400161358.hg.1 ENST00000619235.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400161358.hg.1 NR_107003.1 Refseq
Affymetrix Human Clariom D Assay PSR0400161358.hg.1 ENST00000619235.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400161358.hg.1 NR_107003.1 Refseq
Affymetrix Human Clariom D Assay PSR0400161358.hg.1 TC0400008930.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0