This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04402557_s1
See other MIR7152 GE Assays ›
Gene Symbol:
MIR7152
Gene Name:
microRNA 7152
Gene Aliases:
hsa-mir-7152
Chromosome Location:
Chr.10: 71790747 - 71790800 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04402557_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR7152
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_106974.1 1 25 54

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream chromosome 10 open reading frame 54 C10orf54 B7-H5,
B7H5,
DD1alpha,
GI24,
PD-1H,
PP2135,
SISP1,
VISTA
-17.2kb Anti-sense Chr. 10 - 71747556 - 71773580 on NCBI Build GRCh38
Downstream prosaposin PSAP GLBA,
SAP1
25.5kb Anti-sense Chr. 10 - 71816283 - 71851375 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC1000007966.hg.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000007966.hg.1 ENST00000619512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007966.hg.1 NR_106974.1 Refseq
Affymetrix Human Clariom D Assay PSR1000154111.hg.1 ENST00000619512.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000154111.hg.1 NR_106974.1 Refseq
Affymetrix Human Clariom D Assay PSR1000154111.hg.1 ENST00000619512.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000154111.hg.1 NR_106974.1 Refseq
Affymetrix Human Clariom D Assay PSR1000154111.hg.1 TC1000007966.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000007966.hg.1 ENST00000619512.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007966.hg.1 NR_106974.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0