This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04401791_m1
See other SCHLAP1 GE Assays ›
Gene Symbol:
SCHLAP1
Gene Name:
SWI/SNF complex antagonist associated with prostate cancer 1 (non-protein coding)
Gene Aliases:
LINC00913, PCAT11, PCAT114
Chromosome Location:
Chr.2: 180692104 - 180916939 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04401791_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SCHLAP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_104323.1 2 - 3 421 79
GenBank mRNA JX117422.1 - 2 - 3 421 79

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream CWC22 homolog,
spliceosome-associated protein
CWC22 EIF4GL,
NCM,
fSAPb
-717.3kb Anti-sense Chr. 2 - 179944876 - 180007359 on NCBI Build GRCh38
Downstream ubiquitin conjugating enzyme E2 E3 UBE2E3 UBCH9,
UbcM2
156.3kb Sense Chr. 2 - 180980381 - 181063427 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200010149.hg.1

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 ENST00000629145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 ENST00000629145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 ENST00000629145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 ENST00000629145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 ENST00000629145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 ENST00000629145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104319.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104319.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104319.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104319.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104319.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104319.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104320.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104320.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104320.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104320.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104320.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104321.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104321.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104321.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104321.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104322.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104322.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104322.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104322.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104322.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104322.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104322.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104322.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104323.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104323.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104323.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104323.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104323.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104323.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104323.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104324.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104324.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104324.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104324.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104324.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104324.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104325.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104325.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104325.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104325.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104325.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104325.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010149.hg.1 NR_104325.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0