This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04400024_m1
See other APOBEC3H GE Assays ›
Gene Symbol:
APOBEC3H
Gene Name:
apolipoprotein B mRNA editing enzyme catalytic subunit 3H
Gene Aliases:
A3H, ARP-10, ARP10
Chromosome Location:
Chr.22: 39097204 - 39104067 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04400024_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: APOBEC3H
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001166002.2 NP_001159474.2 4 - 5 669 78
GenBank mRNA BC069023.1 - 4 - 5 654 5441646 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2200007382.hg.1
This assay targets the same location as HTA transcript cluster(s) TC22000311.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

cytidine deamination
negative regulation of transposition
innate immune response
negative regulation of single stranded viral RNA replication via double stranded DNA intermediate
negative regulation of viral process
defense response to virus
DNA cytosine deamination
cytidine deaminase activity
protein binding
zinc ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR22005868.hg.1 ENST00000348946.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22005868.hg.1 ENST00000442487.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22005868.hg.1 NM_001166002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22005868.hg.1 NM_001166003.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22005868.hg.1 NM_181773.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22005868.hg.1 TC22000311.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000348946.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000348946.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000348946.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000348946.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000348946.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000348946.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000348946.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000401756.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000401756.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000401756.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000401756.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000401756.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000401756.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000401756.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000421988.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000421988.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000421988.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000421988.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000421988.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000442487.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000442487.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000442487.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000442487.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000442487.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000442487.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000474235.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000474235.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000474235.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000613677.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000613677.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000613677.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000613677.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000613677.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000613677.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000613996.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000613996.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 ENST00000613996.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_001166002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_001166002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_001166002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_001166002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_001166002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_001166002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_001166002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_001166003.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_001166003.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_001166003.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_001166003.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_001166003.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_001166003.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_001166003.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_001166003.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_001166003.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_001166004.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_001166004.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_001166004.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_001166004.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_001166004.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_001166004.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_181773.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_181773.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_181773.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_181773.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_181773.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_181773.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007382.hg.1 NM_181773.4 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000348946.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000348946.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000348946.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000348946.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000348946.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000348946.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000348946.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000401756.5 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000401756.5 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000401756.5 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000401756.5 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000401756.5 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000401756.5 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000401756.5 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000421988.6 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000421988.6 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000421988.6 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000421988.6 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000421988.6 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000442487.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000442487.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000442487.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000442487.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000442487.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000442487.7 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000474235.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000474235.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000474235.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000613677.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000613677.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000613677.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000613677.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000613677.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000613677.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000613996.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000613996.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 ENST00000613996.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_001166002.2 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_001166002.2 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_001166002.2 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_001166002.2 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_001166002.2 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_001166002.2 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_001166002.2 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_001166003.2 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_001166003.2 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_001166003.2 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_001166003.2 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_001166003.2 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_001166003.2 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_001166003.2 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_001166003.2 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_001166003.2 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_001166004.2 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_001166004.2 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_001166004.2 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_001166004.2 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_001166004.2 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_001166004.2 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_181773.4 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_181773.4 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_181773.4 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_181773.4 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_181773.4 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_181773.4 Refseq
Affymetrix Human Clariom D Assay TC2200007382.hg.1 NM_181773.4 Refseq
Affymetrix Human Clariom D Assay PSR2200152064.hg.1 ENST00000348946.8 Ensembl
Affymetrix Human Clariom D Assay PSR2200152064.hg.1 ENST00000442487.7 Ensembl
Affymetrix Human Clariom D Assay PSR2200152064.hg.1 NM_001166002.2 Refseq
Affymetrix Human Clariom D Assay PSR2200152064.hg.1 NM_001166003.2 Refseq
Affymetrix Human Clariom D Assay PSR2200152064.hg.1 NM_181773.4 Refseq
Affymetrix Human Clariom D Assay PSR2200152064.hg.1 ENST00000348946.8 Ensembl
Affymetrix Human Clariom D Assay PSR2200152064.hg.1 ENST00000442487.7 Ensembl
Affymetrix Human Clariom D Assay PSR2200152064.hg.1 NM_001166002.2 Refseq
Affymetrix Human Clariom D Assay PSR2200152064.hg.1 NM_001166003.2 Refseq
Affymetrix Human Clariom D Assay PSR2200152064.hg.1 NM_181773.4 Refseq
Affymetrix Human Clariom D Assay PSR2200152064.hg.1 TC2200007382.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0