Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009256.hg.1 |
ENST00000589930.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009256.hg.1 |
ENST00000589930.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009256.hg.1 |
ENST00000589930.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009256.hg.1 |
ENST00000589930.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009256.hg.1 |
NR_110763.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009256.hg.1 |
NR_110763.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009256.hg.1 |
NR_110763.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009256.hg.1 |
NR_110763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800163826.hg.1 |
ENST00000589930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800163826.hg.1 |
NR_110763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800163826.hg.1 |
ENST00000589930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800163826.hg.1 |
NR_110763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800163826.hg.1 |
TC1800009256.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1800009256.hg.1 |
ENST00000589930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009256.hg.1 |
ENST00000589930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009256.hg.1 |
ENST00000589930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009256.hg.1 |
ENST00000589930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009256.hg.1 |
NR_110763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009256.hg.1 |
NR_110763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009256.hg.1 |
NR_110763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009256.hg.1 |
NR_110763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800163828.hg.1 |
ENST00000589930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800163828.hg.1 |
NR_110763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800163828.hg.1 |
ENST00000589930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800163828.hg.1 |
NR_110763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800163828.hg.1 |
TC1800009256.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800163825.hg.1 |
ENST00000589930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800163825.hg.1 |
NR_110763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800163825.hg.1 |
ENST00000589930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800163825.hg.1 |
NR_110763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800163825.hg.1 |
TC1800009256.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800163827.hg.1 |
ENST00000589930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800163827.hg.1 |
NR_110763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800163827.hg.1 |
ENST00000589930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800163827.hg.1 |
NR_110763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800163827.hg.1 |
TC1800009256.hg.1 |
Clariom D TC |